missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methylmorpholine N-oxide, 50 wt.% solution in water, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 348270010

 View more versions of this product

Catalog No. AC348270010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Specifications

4-Methylmorpholine N-oxide
7529-22-8,7732-18-5
52.0,50.0
Colorless to Yellow
118.0°C to 119.0°C
Glass bottle
MFCD00005947
07,244
15, 6171
Solubility in water: miscible
C[N+]1([O-])CCOCC1
117.15
CHEBI:52093
Liquid
50 wt.% solution in water
50.0,48.0
-20.0°C
1.1300g/mL
>100°C
C5H11NO2
1 kg
1.13
4-methylmorpholine n-oxide, 4-methylmorpholine 4-oxide, n-methylmorpholine oxide, n-methylmorpholine n-oxide, morpholine, 4-methyl-, 4-oxide, 4-methylmorpholine-4-oxide, methyl morpholine oxide, n-methylmorpholine-n-oxide, nmo, unii-arc64pkj0f
LFTLOKWAGJYHHR-UHFFFAOYSA-N
4-methyl-4-oxidomorpholin-4-ium
82029
117.15
SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only