Learn More
4-Methyloctanoic acid, 98%
CAS: 54947-74-9 | C9H17O2 | 157.23 g/mol
$147.40 - $337.18
Chemical Identifiers
| CAS | 54947-74-9 |
|---|---|
| Molecular Formula | C9H17O2 |
| Molecular Weight (g/mol) | 157.23 |
| MDL Number | MFCD00051938 |
| InChI Key | LEGGANXCVQPIAI-MRVPVSSYSA-M |
| Synonym | octanoic acid, 4-methyl, 4-methyl-octanoic acid, 4-methylcaprylic acid, +/--4-methyloctanoic acid, 4-methyl-n-octanoic acid, 4-methyl octanoic acid, octanoic acid, 4-methyl-, r, fema no. 3575, acmc-20mssw, acmc-20mssx |
| PubChem CID | 62089 |
| IUPAC Name | 4-methyloctanoic acid |
| SMILES | CCCC[C@@H](C)CCC([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2261606
|
Thermo Scientific Chemicals
B2261606 |
5 g |
Each for $147.40
|
|
|||||
|
AAB2261614
|
Thermo Scientific Chemicals
B2261614 |
25 g |
Each for $337.18
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 54947-74-9 | |
| 157.23 | |
| LEGGANXCVQPIAI-MRVPVSSYSA-M | |
| 62089 | |
| CCCC[C@@H](C)CCC([O-])=O |
| C9H17O2 | |
| MFCD00051938 | |
| octanoic acid, 4-methyl, 4-methyl-octanoic acid, 4-methylcaprylic acid, +/--4-methyloctanoic acid, 4-methyl-n-octanoic acid, 4-methyl octanoic acid, octanoic acid, 4-methyl-, r, fema no. 3575, acmc-20mssw, acmc-20mssx | |
| 4-methyloctanoic acid |
Specifications
| 54947-74-9 | |
| 272°C | |
| C9H17O2 | |
| MFCD00051938 | |
| UN3265 | |
| octanoic acid, 4-methyl, 4-methyl-octanoic acid, 4-methylcaprylic acid, +/--4-methyloctanoic acid, 4-methyl-n-octanoic acid, 4-methyl octanoic acid, octanoic acid, 4-methyl-, r, fema no. 3575, acmc-20mssw, acmc-20mssx | |
| CCCC[C@@H](C)CCC([O-])=O | |
| 157.23 | |
| 158.24 | |
| 4-Methyloctanoic acid |
| 0.91 | |
| >110°C (230°F) | |
| 1.4335 | |
| 5 g | |
| 1755426 | |
| LEGGANXCVQPIAI-MRVPVSSYSA-M | |
| 4-methyloctanoic acid | |
| 62089 | |
| 98% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 259-404-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only