Learn More
4-Methylpiperidine, 99%, Thermo Scientific Chemicals
$68.46 - $861.25
Chemical Identifiers
CAS | 626-58-4 |
---|---|
Molecular Formula | C6H13N |
Molecular Weight (g/mol) | 99.18 |
MDL Number | MFCD00006005 |
InChI Key | UZOFELREXGAFOI-UHFFFAOYSA-N |
Synonym | 4-pipecoline, piperidine, 4-methyl, gamma-pipecoline, 4-methylpiperidin, .gamma.-pipecoline, 4-methyl-piperidine, 4-methylpiperdine, 4-methylpyperidine, 4-methyl piperidine, hexahydro-g-picoline |
PubChem CID | 69381 |
IUPAC Name | 4-methylpiperidine |
SMILES | CC1CCNCC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC127511000
|
Thermo Scientific Chemicals
127511000 |
100 g |
Each for $68.46
|
|
|||||
AC127515000
|
Thermo Scientific Chemicals
127515000 |
500 g |
Each for $239.05
|
|
|||||
AC127510025
|
Thermo Scientific Chemicals
127510025 |
2.5 kg |
Each for $861.25
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
626-58-4 | |
99.18 | |
UZOFELREXGAFOI-UHFFFAOYSA-N | |
69381 | |
CC1CCNCC1 |
C6H13N | |
MFCD00006005 | |
4-pipecoline, piperidine, 4-methyl, gamma-pipecoline, 4-methylpiperidin, .gamma.-pipecoline, 4-methyl-piperidine, 4-methylpiperdine, 4-methylpyperidine, 4-methyl piperidine, hexahydro-g-picoline | |
4-methylpiperidine |
Specifications
626-58-4 | |
100.0 | |
Yellow | |
122.0°C | |
Authentic | |
Glass bottle | |
1.4448 to 1.4468 | |
100 g | |
4-pipecoline, piperidine, 4-methyl, gamma-pipecoline, 4-methylpiperidin, .gamma.-pipecoline, 4-methyl-piperidine, 4-methylpiperdine, 4-methylpyperidine, 4-methyl piperidine, hexahydro-g-picoline | |
UZOFELREXGAFOI-UHFFFAOYSA-N | |
4-methylpiperidine | |
69381 | |
99% | |
4-Methylpiperidine, 99% |
98.5 | |
4.0°C to 5.0°C | |
0.8400g/mL | |
13°C | |
98.5% min. (GC) | |
C6H13N | |
MFCD00006005 | |
0.84 | |
Solubility in water: miscible. | |
CC1CCNCC1 | |
99.18 | |
99.18 | |
Liquid |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic in contact with skin.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
GHS Signal Word: Danger
EINECSNumber : 210-954-1
TSCA : TSCA
RUO – Research Use Only