missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methylstyrene (stabilized with TBC) 96.0+%, TCI America™

Supplier:  TCI America M0428100ML

 View more versions of this product

Catalog No. M0428100ML


Explore available promotions

Description

Description

Specifications

Specifications

4-Methylstyrene (stabilized with TBC)
Yellow
C9H10
100 mL
4-methylstyrene, 1-methyl-4-vinylbenzene, p-methylstyrene, 4-vinyltoluene, p-vinyltoluene, benzene, 1-ethenyl-4-methyl, styrene, p-methyl, p-methyl styrene, 1-p-tolylethene, para-methylstyrene
CC1=CC=C(C=C)C=C1
118.18
118.18
Liquid
622-97-9
175°C
MFCD00008621
2618
JLBJTVDPSNHSKJ-UHFFFAOYSA-N
1-ethenyl-4-methylbenzene
12161
≥96.0% (GC)

Chemical Identifiers

622-97-9
118.18
JLBJTVDPSNHSKJ-UHFFFAOYSA-N
12161
CC1=CC=C(C=C)C=C1
C9H10
MFCD00008621
4-methylstyrene, 1-methyl-4-vinylbenzene, p-methylstyrene, 4-vinyltoluene, p-vinyltoluene, benzene, 1-ethenyl-4-methyl, styrene, p-methyl, p-methyl styrene, 1-p-tolylethene, para-methylstyrene
1-ethenyl-4-methylbenzene
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0008

RTECSNumber : WL5076000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions