missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Methylstyrene (stabilized with TBC) 96.0+%, TCI America™
Supplier: TCI America M0428500ML
Specifications
4-Methylstyrene (stabilized with TBC) | |
Yellow | |
C9H10 | |
500 mL | |
4-methylstyrene, 1-methyl-4-vinylbenzene, p-methylstyrene, 4-vinyltoluene, p-vinyltoluene, benzene, 1-ethenyl-4-methyl, styrene, p-methyl, p-methyl styrene, 1-p-tolylethene, para-methylstyrene | |
CC1=CC=C(C=C)C=C1 | |
118.18 | |
118.18 | |
Liquid |
622-97-9 | |
175°C | |
MFCD00008621 | |
2618 | |
JLBJTVDPSNHSKJ-UHFFFAOYSA-N | |
1-ethenyl-4-methylbenzene | |
12161 | |
≥96.0% (GC) |
Chemical Identifiers
622-97-9 | |
118.18 | |
JLBJTVDPSNHSKJ-UHFFFAOYSA-N | |
12161 | |
CC1=CC=C(C=C)C=C1 |
C9H10 | |
MFCD00008621 | |
4-methylstyrene, 1-methyl-4-vinylbenzene, p-methylstyrene, 4-vinyltoluene, p-vinyltoluene, benzene, 1-ethenyl-4-methyl, styrene, p-methyl, p-methyl styrene, 1-p-tolylethene, para-methylstyrene | |
1-ethenyl-4-methylbenzene |
Safety and Handling
EINECSNumber : (3)-0008
RTECSNumber : WL5076000
TSCA : Yes
Recommended Storage : Refrigerator