missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylsulfonyl)aniline 98.0+%, TCI America™

$95.06 - $374.68

Chemical Identifiers

CAS 5470-49-5
Molecular Formula C7H9NO2S
Molecular Weight (g/mol) 171.21
MDL Number MFCD00025353
InChI Key XJEVFFNOMKXBLU-UHFFFAOYSA-N
Synonym 4-methylsulfonyl aniline, 4-methylsulphonyl aniline, 4-mesylaniline, 4-methanesulfonylaniline, p-methylsulfonyl aniline, benzeneamine, 4-methylsulfonyl, benzenamine, 4-methylsulfonyl, 4-methylsulfonyl benzenamine, 4-methylsulphonylaniline
PubChem CID 79617
IUPAC Name 4-methanesulfonylaniline
SMILES CS(=O)(=O)C1=CC=C(N)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A272525G
View Documents
TCI America
A272525G
25 g
Each for $95.06
Only null left
 
A2725100G
View Documents
TCI America
A2725100G
100 g
Each for $374.68
Only null left
 
Description

Description

Specifications

Chemical Identifiers

5470-49-5
171.21
XJEVFFNOMKXBLU-UHFFFAOYSA-N
79617
CS(=O)(=O)C1=CC=C(N)C=C1
C7H9NO2S
MFCD00025353
4-methylsulfonyl aniline, 4-methylsulphonyl aniline, 4-mesylaniline, 4-methanesulfonylaniline, p-methylsulfonyl aniline, benzeneamine, 4-methylsulfonyl, benzenamine, 4-methylsulfonyl, 4-methylsulfonyl benzenamine, 4-methylsulphonylaniline
4-methanesulfonylaniline

Specifications

5470-49-5
White
MFCD00025353
4-methylsulfonyl aniline, 4-methylsulphonyl aniline, 4-mesylaniline, 4-methanesulfonylaniline, p-methylsulfonyl aniline, benzeneamine, 4-methylsulfonyl, benzenamine, 4-methylsulfonyl, 4-methylsulfonyl benzenamine, 4-methylsulphonylaniline
CS(=O)(=O)C1=CC=C(N)C=C1
171.21
171.21
Crystalline Powder
137°C
C7H9NO2S
25 g
XJEVFFNOMKXBLU-UHFFFAOYSA-N
4-methanesulfonylaniline
79617
≥98.0% (GC,T)
4-(Methylsulfonyl)aniline
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2137

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'