missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methylumbelliferyl beta-D-glucopyranoside, 99%, Thermo Scientific Chemicals

$82.53 - $843.77

Chemical Identifiers

CAS 18997-57-4
Molecular Formula C16H18O8
Molecular Weight (g/mol) 338.312
MDL Number MFCD00036773
InChI Key YUDPTGPSBJVHCN-YMILTQATSA-N
Synonym 4-methylumbelliferyl, a-d-glucoside, glc1-b-4mu, 7-, a-d-glucopyranosyloxy-4-methyl-2h-1-benzopyran-2-one, 4-methylumbelliferyl glucopyranoside, 4-methyl-7-2s,4s,5s-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy chromen-2-one
PubChem CID 2733779
IUPAC Name 4-methyl-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one
SMILES CC1=CC(=O)OC2=C1C=CC(=C2)OC3C(C(C(C(O3)CO)O)O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66630MD
View Documents Promotion Details
Thermo Scientific Chemicals
J66630MD
250 mg
Each for $82.53
 
AAJ6663003
View Documents Promotion Details
Thermo Scientific Chemicals
J6663003
1 g
Each for $238.32
 
AAJ6663006
View Documents Promotion Details
Thermo Scientific Chemicals
J6663006
5 g
Each for $843.77
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

18997-57-4
338.312
YUDPTGPSBJVHCN-YMILTQATSA-N
2733779
CC1=CC(=O)OC2=C1C=CC(=C2)OC3C(C(C(C(O3)CO)O)O)O
C16H18O8
MFCD00036773
4-methylumbelliferyl, a-d-glucoside, glc1-b-4mu, 7-, a-d-glucopyranosyloxy-4-methyl-2h-1-benzopyran-2-one, 4-methylumbelliferyl glucopyranoside, 4-methyl-7-2s,4s,5s-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy chromen-2-one
4-methyl-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one

Specifications

18997-57-4
White
MFCD00036773
94673
Soluble in DMF or water
CC1=CC(=O)OC2=C1C=CC(=C2)OC3C(C(C(C(O3)CO)O)O)O
338.312
338.31
Powder
210°C to 212°C
C16H18O8
250 mg
4-methylumbelliferyl, a-d-glucoside, glc1-b-4mu, 7-, a-d-glucopyranosyloxy-4-methyl-2h-1-benzopyran-2-one, 4-methylumbelliferyl glucopyranoside, 4-methyl-7-2s,4s,5s-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy chromen-2-one
YUDPTGPSBJVHCN-YMILTQATSA-N
4-methyl-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one
2733779
99%
4-Methylumbelliferyl beta-D-glucopyranoside
Safety and Handling

Safety and Handling

EINECSNumber : 242-736-7

TSCA : No

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only