missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methylumbelliferyl-beta-D-glucuronide, 98%, Thermo Scientific Chemicals

$130.42 - $479.92

Chemical Identifiers

CAS 6160-80-1
Molecular Formula C16H16O9
Molecular Weight (g/mol) 352.295
MDL Number MFCD00036772
InChI Key ARQXEQLMMNGFDU-JHZZJYKESA-N
Synonym 4-methylumbelliferyl-beta-d-glucuronide, 4-methylumbelliferyl glucuronide, 4-methylumbelliferone glucuronide, mug trihydrate, 4-methylumbelliferyl beta-glucuronide, 4-methyl-2-oxo-2h-1-benzopyran-7-yl beta-d-glucopyranosiduronic acid, 2s,3s,4s,5r,6s-3,4,5-trihydroxy-6-4-methyl-2-oxo-2h-chromen-7-yl oxy tetrahydro-2h-pyran-2-carboxylic acid, 2s,3s,4s,5r,6s-3,4,5-trihydroxy-6-4-methyl-2-oxo-chromen-7-yl oxy-tetrahydropyran-2-carboxylic acid, 2s,3s,4s,6s,5r-3,4,5-trihydroxy-6-4-methyl-2-oxochromen-7-yloxy-2h-3,4,5,6-tetrahydropyran-2-carboxylic acid, 2s,3s,4s,5r,6s-3,4,5-trihydroxy-6-4-methyl-2-oxochromen-7-yl oxy oxane-2-carboxylic acid
PubChem CID 91553
ChEBI CHEBI:1904
IUPAC Name (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(4-methyl-2-oxochromen-7-yl)oxyoxane-2-carboxylic acid
SMILES CC1=CC(=O)OC2=C1C=CC(=C2)OC3C(C(C(C(O3)C(=O)O)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB21190MF
View Documents
Thermo Scientific Chemicals
B21190MF
50 mg
Each for $130.42
 
AAB21190MD
View Documents
Thermo Scientific Chemicals
B21190MD
250 mg
Each for $479.92
 
Description

Description

It is a fluorogenic substrate for measuring of β-glucoronidiase activity, for rapid sensitive identification of Escherichia coli. MUG is a substrate for glucuronidase. It is ideal for plant molecular study as a way to examine gene expression under various promoters. And while it works well for measuring activity in plant lysate and leaf disks, it can also work in whole plant assays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a fluorogenic substrate for measuring of ß-glucoronidiase activity, for rapid sensitive identification of Escherichia coli. MUG is a substrate for glucuronidase. It is ideal for plant molecular study as a way to examine gene expression under various promoters. And while it works well for measuring activity in plant lysate and leaf disks, it can also work in whole plant assays.

Solubility
Soluble in hot water.

Notes
Light sensitive. Store away from alkali metals bases and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in freezer at -20°C.
Specifications

Chemical Identifiers

6160-80-1
352.295
ARQXEQLMMNGFDU-JHZZJYKESA-N
91553
(2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(4-methyl-2-oxochromen-7-yl)oxyoxane-2-carboxylic acid
C16H16O9
MFCD00036772
4-methylumbelliferyl-beta-d-glucuronide, 4-methylumbelliferyl glucuronide, 4-methylumbelliferone glucuronide, mug trihydrate, 4-methylumbelliferyl beta-glucuronide, 4-methyl-2-oxo-2h-1-benzopyran-7-yl beta-d-glucopyranosiduronic acid, 2s,3s,4s,5r,6s-3,4,5-trihydroxy-6-4-methyl-2-oxo-2h-chromen-7-yl oxy tetrahydro-2h-pyran-2-carboxylic acid, 2s,3s,4s,5r,6s-3,4,5-trihydroxy-6-4-methyl-2-oxo-chromen-7-yl oxy-tetrahydropyran-2-carboxylic acid, 2s,3s,4s,6s,5r-3,4,5-trihydroxy-6-4-methyl-2-oxochromen-7-yloxy-2h-3,4,5,6-tetrahydropyran-2-carboxylic acid, 2s,3s,4s,5r,6s-3,4,5-trihydroxy-6-4-methyl-2-oxochromen-7-yl oxy oxane-2-carboxylic acid
CHEBI:1904
CC1=CC(=O)OC2=C1C=CC(=C2)OC3C(C(C(C(O3)C(=O)O)O)O)O

Specifications

6160-80-1
MFCD00036772
Light sensitive
Soluble in hot water.
−106° (c=0.1 in Water)
(2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(4-methyl-2-oxochromen-7-yl)oxyoxane-2-carboxylic acid
91553
352.3
4-Methylumbelliferyl-beta-D-glucuronide
C16H16O9
50 mg
4-methylumbelliferyl-beta-d-glucuronide, 4-methylumbelliferyl glucuronide, 4-methylumbelliferone glucuronide, mug trihydrate, 4-methylumbelliferyl beta-glucuronide, 4-methyl-2-oxo-2h-1-benzopyran-7-yl beta-d-glucopyranosiduronic acid, 2s,3s,4s,5r,6s-3,4,5-trihydroxy-6-4-methyl-2-oxo-2h-chromen-7-yl oxy tetrahydro-2h-pyran-2-carboxylic acid, 2s,3s,4s,5r,6s-3,4,5-trihydroxy-6-4-methyl-2-oxo-chromen-7-yl oxy-tetrahydropyran-2-carboxylic acid, 2s,3s,4s,6s,5r-3,4,5-trihydroxy-6-4-methyl-2-oxochromen-7-yloxy-2h-3,4,5,6-tetrahydropyran-2-carboxylic acid, 2s,3s,4s,5r,6s-3,4,5-trihydroxy-6-4-methyl-2-oxochromen-7-yl oxy oxane-2-carboxylic acid
ARQXEQLMMNGFDU-JHZZJYKESA-N
CC1=CC(=O)OC2=C1C=CC(=C2)OC3C(C(C(C(O3)C(=O)O)O)O)O
352.295
CHEBI:1904
98%
Videos
Safety and Handling

Safety and Handling

H500

EINECSNumber : 228-186-0

TSCA : Yes

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only