missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methylvaleric Acid 97.0+%, TCI America™
$154.71 - $154.71
Chemical Identifiers
| CAS | 646-07-1 |
|---|---|
| Molecular Formula | C6H12O2 |
| Molecular Weight (g/mol) | 116.16 |
| MDL Number | MFCD00002803 |
| InChI Key | FGKJLKRYENPLQH-UHFFFAOYSA-N |
| Synonym | 4-methylvaleric acid, isocaproic acid, isohexanoic acid, pentanoic acid, 4-methyl, isobutylacetic acid, 4-methyl-pentanoic acid, valeric acid, 4-methyl, 4-methyl valeric acid, isocaproate, 4,4-dimethylbutanoic acid |
| PubChem CID | 12587 |
| ChEBI | CHEBI:74903 |
| IUPAC Name | 4-methylpentanoic acid |
| SMILES | CC(C)CCC(=O)O |
Chemical Identifiers
| 646-07-1 | |
| 116.16 | |
| FGKJLKRYENPLQH-UHFFFAOYSA-N | |
| 12587 | |
| 4-methylpentanoic acid |
| C6H12O2 | |
| MFCD00002803 | |
| 4-methylvaleric acid, isocaproic acid, isohexanoic acid, pentanoic acid, 4-methyl, isobutylacetic acid, 4-methyl-pentanoic acid, valeric acid, 4-methyl, 4-methyl valeric acid, isocaproate, 4,4-dimethylbutanoic acid | |
| CHEBI:74903 | |
| CC(C)CCC(=O)O |
Specifications
| 646-07-1 | |
| 201°C | |
| MFCD00002803 | |
| 2810 | |
| FGKJLKRYENPLQH-UHFFFAOYSA-N | |
| 4-methylpentanoic acid | |
| 12587 | |
| 116.16 | |
| Liquid |
| Yellow | |
| C6H12O2 | |
| 25 mL | |
| 4-methylvaleric acid, isocaproic acid, isohexanoic acid, pentanoic acid, 4-methyl, isobutylacetic acid, 4-methyl-pentanoic acid, valeric acid, 4-methyl, 4-methyl valeric acid, isocaproate, 4,4-dimethylbutanoic acid | |
| CC(C)CCC(=O)O | |
| 116.16 | |
| CHEBI:74903 | |
| ≥97.0% (GC) | |
| 4-Methylvaleric Acid |
Safety and Handling
EINECSNumber : (2)-0608
RTECSNumber : NR2975000
TSCA : Yes