missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-n-Butyl-4'-cyanobiphenyl, 98%, Thermo Scientific™

$56.93 - $191.92

Chemical Identifiers

CAS 52709-83-8
Molecular Formula C17H17N
Molecular Weight (g/mol) 235.33
MDL Number MFCD00506009
InChI Key PJPLBHHDTUICNN-UHFFFAOYSA-N
Synonym 4'-butyl-1,1'-biphenyl-4-carbonitrile, 4-butyl-4'-cyanobiphenyl, 4'-butyl-4-biphenylcarbonitrile, 4-cyano-4'-butylbiphenyl, 1,1'-biphenyl-4-carbonitrile, 4'-butyl, 4-n-butyl-4'-cyanobiphenyl, 4-4-butylphenyl benzonitrile, 4'-butyl 1,1'-biphenyl-4-carbonitrile, 4'-butyl-4-cyanobiphenyl
PubChem CID 104288
IUPAC Name 4-(4-butylphenyl)benzonitrile
SMILES CCCCC1=CC=C(C=C1)C2=CC=C(C=C2)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2233803
View Documents
Thermo Scientific Chemicals
B2233803
1 g
Each for $56.93
 
AAB2233806
View Documents
Thermo Scientific Chemicals
B2233806
5 g
Each for $191.92
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

52709-83-8
235.33
PJPLBHHDTUICNN-UHFFFAOYSA-N
104288
CCCCC1=CC=C(C=C1)C2=CC=C(C=C2)C#N
C17H17N
MFCD00506009
4'-butyl-1,1'-biphenyl-4-carbonitrile, 4-butyl-4'-cyanobiphenyl, 4'-butyl-4-biphenylcarbonitrile, 4-cyano-4'-butylbiphenyl, 1,1'-biphenyl-4-carbonitrile, 4'-butyl, 4-n-butyl-4'-cyanobiphenyl, 4-4-butylphenyl benzonitrile, 4'-butyl 1,1'-biphenyl-4-carbonitrile, 4'-butyl-4-cyanobiphenyl
4-(4-butylphenyl)benzonitrile

Specifications

52709-83-8
210°C to 212°C (2 mmHg)
MFCD00506009
UN3439
PJPLBHHDTUICNN-UHFFFAOYSA-N
4-(4-butylphenyl)benzonitrile
104288
98%
49°C to 51°C
C17H17N
1 g
4'-butyl-1,1'-biphenyl-4-carbonitrile, 4-butyl-4'-cyanobiphenyl, 4'-butyl-4-biphenylcarbonitrile, 4-cyano-4'-butylbiphenyl, 1,1'-biphenyl-4-carbonitrile, 4'-butyl, 4-n-butyl-4'-cyanobiphenyl, 4-4-butylphenyl benzonitrile, 4'-butyl 1,1'-biphenyl-4-carbonitrile, 4'-butyl-4-cyanobiphenyl
CCCCC1=CC=C(C=C1)C2=CC=C(C=C2)C#N
235.33
235.33
4-n-Butyl-4'-cyanobiphenyl
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H304-H312
Toxic if inhaled.
May be fatal if swallowed and enters airways.
Harmful in contact with skin.

P261-P271-P280-P301+P310-P302+P352-P304+P340-P312-P331-P363-P501c

H304-H312+H332

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

EINECSNumber : 258-119-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'