missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Nitro-1,2-phenylenediamine Monohydrochloride 99.0+%, TCI America™
Supplier: TCI America A50815G
Specifications
4-Nitro-1,2-phenylenediamine Monohydrochloride [Sensitive reagent for the determination of Se by GC- | |
C6H8ClN3O2 | |
5 g | |
IFTLGFVLGVXRBT-UHFFFAOYSA-N | |
4-nitrobenzene-1,2-diamine;hydrochloride | |
5743500 | |
≥99.0% (T) |
53209-19-1 | |
MFCD00043273 | |
1,2-Diamino-4-nitrobenzene Monohydrochloride | |
C1=CC(=C(C=C1[N+](=O)[O-])N)N.Cl | |
189.599 | |
189.60 | |
Crystalline Powder |
Chemical Identifiers
53209-19-1 | |
189.599 | |
IFTLGFVLGVXRBT-UHFFFAOYSA-N | |
5743500 | |
C1=CC(=C(C=C1[N+](=O)[O-])N)N.Cl |
C6H8ClN3O2 | |
MFCD00043273 | |
1,2-Diamino-4-nitrobenzene Monohydrochloride | |
4-nitrobenzene-1,2-diamine;hydrochloride |
Safety and Handling
EINECSNumber : (3)-0402
TSCA : No