missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitro-D-phenylalanine, 98%, Thermo Scientific Chemicals
$86.86 - $86.86
Chemical Identifiers
| CAS | 56613-61-7 |
|---|---|
| Molecular Formula | C9H10N2O4 |
| Molecular Weight (g/mol) | 210.19 |
| MDL Number | MFCD00066016 |
| InChI Key | GTVVZTAFGPQSPC-UHFFFAOYNA-N |
| Synonym | 4-nitro-d-phenylalanine, h-d-phe 4-no2-oh, p-nitro-d-phenylalanine, 2r-2-amino-3-4-nitrophenyl propanoic acid, d-phenylalanine, 4-nitro, r-2-amino-3-4-nitrophenyl propanoic acid, d-4-nitrophe, ambotzhaa1044, pubchem13021, d-4-nitro phenylalanine |
| PubChem CID | 679497 |
| IUPAC Name | 2-amino-3-(4-nitrophenyl)propanoic acid |
| SMILES | NC(CC1=CC=C(C=C1)[N+]([O-])=O)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2695903
|
Thermo Scientific Chemicals
H2695903 |
1 g |
Each for $86.86
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 56613-61-7 | |
| 210.19 | |
| GTVVZTAFGPQSPC-UHFFFAOYNA-N | |
| 679497 | |
| NC(CC1=CC=C(C=C1)[N+]([O-])=O)C(O)=O |
| C9H10N2O4 | |
| MFCD00066016 | |
| 4-nitro-d-phenylalanine, h-d-phe 4-no2-oh, p-nitro-d-phenylalanine, 2r-2-amino-3-4-nitrophenyl propanoic acid, d-phenylalanine, 4-nitro, r-2-amino-3-4-nitrophenyl propanoic acid, d-4-nitrophe, ambotzhaa1044, pubchem13021, d-4-nitro phenylalanine | |
| 2-amino-3-(4-nitrophenyl)propanoic acid |
Specifications
| 56613-61-7 | |
| MFCD00066016 | |
| GTVVZTAFGPQSPC-UHFFFAOYNA-N | |
| 2-amino-3-(4-nitrophenyl)propanoic acid | |
| 210.19 | |
| 210.19 | |
| 4-Nitro-D-phenylalanine |
| C9H10N2O4 | |
| 4-nitro-d-phenylalanine, h-d-phe 4-no2-oh, p-nitro-d-phenylalanine, 2r-2-amino-3-4-nitrophenyl propanoic acid, d-phenylalanine, 4-nitro, r-2-amino-3-4-nitrophenyl propanoic acid, d-4-nitrophe, ambotzhaa1044, pubchem13021, d-4-nitro phenylalanine | |
| NC(CC1=CC=C(C=C1)[N+]([O-])=O)C(O)=O | |
| 1 g | |
| 679497 | |
| 98% |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 213-446-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only