missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitro-o-phenylenediamine, 97%, Thermo Scientific Chemicals

$122.52 - $427.33

Chemical Identifiers

CAS 99-56-9
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007724
InChI Key RAUWPNXIALNKQM-UHFFFAOYSA-N
Synonym 4-nitro-o-phenylenediamine, 1,2-diamino-4-nitrobenzene, 4-nitro-1,2-phenylenediamine, 2-amino-4-nitroaniline, 3,4-diaminonitrobenzene, p-nitro-o-phenylenediamine, 1,2-benzenediamine, 4-nitro, 4-nitro-1,2-diaminobenzene, 4-nitrophenylenediamine, 4-nitro-1,2-benzenediamine
PubChem CID 5111791
ChEBI CHEBI:67116
IUPAC Name 4-nitrobenzene-1,2-diamine
SMILES NC1=CC=C(C=C1N)[N+]([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0469322
View Documents
Thermo Scientific Chemicals
L0469322
100 g
Each for $122.52
 
AAL0469336
View Documents
Thermo Scientific Chemicals
L0469336
500 g
Each for $427.33
 
Description

Description

4-Nitro-o-phenylenediamine is used to study the in vitro activation of 4-nitro-o-phenylenediamine into a potent mutagen in Salmonella typhimurium strain TA98

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitro-o-phenylenediamine is used to study the in vitro activation of 4-nitro-o-phenylenediamine into a potent mutagen in Salmonella typhimurium strain TA98

Solubility
Slightly soluble in water (1.2 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

99-56-9
153.14
RAUWPNXIALNKQM-UHFFFAOYSA-N
5111791
4-nitrobenzene-1,2-diamine
C6H7N3O2
MFCD00007724
4-nitro-o-phenylenediamine, 1,2-diamino-4-nitrobenzene, 4-nitro-1,2-phenylenediamine, 2-amino-4-nitroaniline, 3,4-diaminonitrobenzene, p-nitro-o-phenylenediamine, 1,2-benzenediamine, 4-nitro, 4-nitro-1,2-diaminobenzene, 4-nitrophenylenediamine, 4-nitro-1,2-benzenediamine
CHEBI:67116
NC1=CC=C(C=C1N)[N+]([O-])=O

Specifications

99-56-9
C6H7N3O2
100 g
14,6623
Slightly soluble in water (1.2g/L at 20°C).
NC1=CC=C(C=C1N)[N+]([O-])=O
153.14
CHEBI:67116
97%
199°C to 203°C
MFCD00007724
608106
4-nitro-o-phenylenediamine, 1,2-diamino-4-nitrobenzene, 4-nitro-1,2-phenylenediamine, 2-amino-4-nitroaniline, 3,4-diaminonitrobenzene, p-nitro-o-phenylenediamine, 1,2-benzenediamine, 4-nitro, 4-nitro-1,2-diaminobenzene, 4-nitrophenylenediamine, 4-nitro-1,2-benzenediamine
RAUWPNXIALNKQM-UHFFFAOYSA-N
4-nitrobenzene-1,2-diamine
5111791
153.14
4-Nitro-o-phenylenediamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H341-H317
Toxic if swallowed.
Suspected of causing genetic defects.
May cause an allergic skin reaction.

P201-P202-P261-P264b-P270-P272-P280g-P281-P301+P312-P302+P352-P308+P313-P330-P333+P313-P363-P501c

H302-H317-H340

EINECSNumber : 202-766-3

RTECSNumber : ST2975000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only