missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitroanisole, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 147022500

 View more versions of this product

Catalog No. AC147022500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Nitroanisole
100-17-4
100.0
Brown to Green
260.0°C
Authentic
Glass bottle
O2NC6H4OCH3
250 g
1.23
4-nitroanisole, p-nitroanisole, 4-methoxynitrobenzene, p-methoxynitrobenzene, benzene, 1-methoxy-4-nitro, nitroanisole, p-nitroanisol, anisole, p-nitro, 4-nitrophenyl methyl ether, unii-g989z7wolh
BNUHAJGCKIQFGE-UHFFFAOYSA-N
1-methoxy-4-nitrobenzene
7485
153.14
Crystals
99+%
99.0
51.0°C to 53.0°C
1.2300g/mL
130°C
99% min. (GC)
C7H7NO3
MFCD00007327
06, 230
15, 6671
Solubility in water: 0.468g/L (20°C). Other solubilities: soluble in alcohol and ether
COC1=CC=C(C=C1)[N+]([O-])=O
153.14
CHEBI:1911
99+%

Chemical Identifiers

100-17-4
153.14
BNUHAJGCKIQFGE-UHFFFAOYSA-N
7485
1-methoxy-4-nitrobenzene
C7H7NO3
MFCD00007327
4-nitroanisole, p-nitroanisole, 4-methoxynitrobenzene, p-methoxynitrobenzene, benzene, 1-methoxy-4-nitro, nitroanisole, p-nitroanisol, anisole, p-nitro, 4-nitrophenyl methyl ether, unii-g989z7wolh
CHEBI:1911
COC1=CC=C(C=C1)[N+]([O-])=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Suspected of causing genetic defects.

GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-825-3

RTECSNumber : BZ8800000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only