Learn More
4-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals
$73.26 - $949.27
Chemical Identifiers
CAS | 555-16-8 |
---|---|
Molecular Formula | C7H5NO3 |
Molecular Weight (g/mol) | 151.12 |
MDL Number | MFCD00007346 |
InChI Key | BXRFQSNOROATLV-UHFFFAOYSA-N |
Synonym | p-nitrobenzaldehyde, benzaldehyde, 4-nitro, p-formylnitrobenzene, benzaldehyde, p-nitro, 4-nitrobenzaldehydde, 4-nitro-benzaldehyde, 4-formylnitrobenzene, p-nitro benzaldehyde, 4-nitro benzaldehyde, ccris 1675 |
PubChem CID | 541 |
ChEBI | CHEBI:66926 |
IUPAC Name | 4-nitrobenzaldehyde |
SMILES | [O-][N+](=O)C1=CC=C(C=O)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1165514
|
Thermo Scientific Chemicals
A1165514 |
25 g |
Each for $73.26
|
|
|||||
AAA1165522
|
Thermo Scientific Chemicals
A1165522 |
100 g |
Each for $209.70
|
|
|||||
AAA1165536
|
Thermo Scientific Chemicals
A1165536 |
500 g |
Each for $949.27
|
|
|||||
Description
4-Nitrobenzaldehyde is used in the preparation of homoallylic alcohols. It is also involved in the development and evaluation of a series of tripeptide organocatalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Nitrobenzaldehyde is used in the preparation of homoallylic alcohols. It is also involved in the development and evaluation of a series of tripeptide organocatalysts.
Solubility
Soluble in water, ethanol, benzene, glacial acetic acid. Slightly soluble in ether.
Notes
Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
Chemical Identifiers
555-16-8 | |
151.12 | |
BXRFQSNOROATLV-UHFFFAOYSA-N | |
541 | |
4-nitrobenzaldehyde |
C7H5NO3 | |
MFCD00007346 | |
p-nitrobenzaldehyde, benzaldehyde, 4-nitro, p-formylnitrobenzene, benzaldehyde, p-nitro, 4-nitrobenzaldehydde, 4-nitro-benzaldehyde, 4-formylnitrobenzene, p-nitro benzaldehyde, 4-nitro benzaldehyde, ccris 1675 | |
CHEBI:66926 | |
[O-][N+](=O)C1=CC=C(C=O)C=C1 |
Specifications
555-16-8 | |
C7H5NO3 | |
25 g | |
Air Sensitive | |
p-nitrobenzaldehyde, benzaldehyde, 4-nitro, p-formylnitrobenzene, benzaldehyde, p-nitro, 4-nitrobenzaldehydde, 4-nitro-benzaldehyde, 4-formylnitrobenzene, p-nitro benzaldehyde, 4-nitro benzaldehyde, ccris 1675 | |
BXRFQSNOROATLV-UHFFFAOYSA-N | |
4-nitrobenzaldehyde | |
541 | |
151.12 | |
4-Nitrobenzaldehyde |
104°C to 106°C | |
MFCD00007346 | |
386796 | |
14,6587 | |
Soluble in water,ethanol,benzene,glacial acetic acid. Slightly soluble in ether. | |
[O-][N+](=O)C1=CC=C(C=O)C=C1 | |
151.12 | |
CHEBI:66926 | |
99% |
Safety and Handling
GHS H Statement
H319-H317
Causes serious eye irritation.
May cause an allergic skin reaction.
P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P363-P501c
H317-H319
EINECSNumber : 209-084-5
RTECSNumber : CU7350000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only