Learn More
4-Nitrobenzenesulfonyl chloride, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 415851000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Nitrobenzenesulfonyl chloride | |
98-74-8 | |
100.0 | |
Yellow to Brown | |
Authentic | |
Glass bottle | |
SO2ClC6H4NO2 | |
100 g | |
4-nitrobenzenesulfonylchloride, 4-nitrobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-nitro, 4-nitrobenzenesulphonyl chloride, 4-nitrophenylsulfonyl chloride, p-nitrophenylsulfonyl chloride, 4-nitrobenzenesulfonic acid chloride, p-nitrobenzenesulfonyl chloride, 4-nitrobenzenesulphonylchloride, 4-nitro-benzenesulfonyl chloride | |
JXRGUPLJCCDGKG-UHFFFAOYSA-N | |
4-nitrobenzenesulfonyl chloride | |
7404 | |
95% |
95% | |
94.0 | |
75°C to 80°C | |
143°C to 144°C (2.0 hPa) | |
94% min. (GC) | |
C6H4ClNO4S | |
MFCD00007445 | |
11, 72 | |
Solubility in water: insoluble | |
[O-][N+](=O)C1=CC=C(C=C1)S(Cl)(=O)=O | |
221.61 | |
221.62 | |
Powder |
Chemical Identifiers
98-74-8 | |
221.61 | |
JXRGUPLJCCDGKG-UHFFFAOYSA-N | |
7404 | |
[O-][N+](=O)C1=CC=C(C=C1)S(Cl)(=O)=O |
C6H4ClNO4S | |
MFCD00007445 | |
4-nitrobenzenesulfonylchloride, 4-nitrobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-nitro, 4-nitrobenzenesulphonyl chloride, 4-nitrophenylsulfonyl chloride, p-nitrophenylsulfonyl chloride, 4-nitrobenzenesulfonic acid chloride, p-nitrobenzenesulfonyl chloride, 4-nitrobenzenesulphonylchloride, 4-nitro-benzenesulfonyl chloride | |
4-nitrobenzenesulfonyl chloride |
Safety and Handling
GHS H Statement
Harmful to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 202-697-9
TSCA : TSCA
RUO – Research Use Only