missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Nitrobenzenethiol 95.0+%, TCI America™
$112.57 - $383.14
Chemical Identifiers
CAS | 1849-36-1 |
---|---|
Molecular Formula | C6H5NO2S |
Molecular Weight (g/mol) | 155.171 |
MDL Number | MFCD00007343 |
InChI Key | AXBVSRMHOPMXBA-UHFFFAOYSA-N |
Synonym | 4-nitrothiophenol, 4-nitrobenzenethiol, p-nitrothiophenol, benzenethiol, 4-nitro, p-nitrophenyl mercaptan, p-nitrobenzenethiol, benzenethiol, p-nitro, 4-nitrobenzene-1-thiol, 4-nitrothiophenolate, 4-nitro-benzenethiol |
PubChem CID | 15809 |
SMILES | C1=CC(=CC=C1[N+](=O)[O-])S |
Chemical Identifiers
1849-36-1 | |
155.171 | |
AXBVSRMHOPMXBA-UHFFFAOYSA-N | |
15809 |
C6H5NO2S | |
MFCD00007343 | |
4-nitrothiophenol, 4-nitrobenzenethiol, p-nitrothiophenol, benzenethiol, 4-nitro, p-nitrophenyl mercaptan, p-nitrobenzenethiol, benzenethiol, p-nitro, 4-nitrobenzene-1-thiol, 4-nitrothiophenolate, 4-nitro-benzenethiol | |
C1=CC(=CC=C1[N+](=O)[O-])S |
Specifications
1849-36-1 | |
C6H5NO2S | |
5 g | |
AXBVSRMHOPMXBA-UHFFFAOYSA-N | |
155.171 | |
155.17 | |
Crystalline Powder |
79°C | |
MFCD00007343 | |
4-nitrothiophenol, 4-nitrobenzenethiol, p-nitrothiophenol, benzenethiol, 4-nitro, p-nitrophenyl mercaptan, p-nitrobenzenethiol, benzenethiol, p-nitro, 4-nitrobenzene-1-thiol, 4-nitrothiophenolate, 4-nitro-benzenethiol | |
C1=CC(=CC=C1[N+](=O)[O-])S | |
15809 | |
≥95.0% (GC,T) | |
4-Nitrobenzenethiol |
Safety and Handling
RTECSNumber : DC1850000
TSCA : No