missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitrobenzylamine hydrochloride, 94%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2174806

 View more versions of this product

Catalog No. AAB2174806


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Nitrobenzylamine hydrochloride
255°C to 257°C (decomposition)
MFCD00012863
3629994
SMIXZZMSWYOQPW-UHFFFAOYSA-N
(4-nitrophenyl)methanamine;hydrochloride
11252467
97%
18600-42-5
C7H9ClN2O2
5 g
4-nitrobenzylamine hydrochloride, 4-nitrophenyl methanamine hydrochloride, p-nitrobenzylamine hydrochloride, 4-nitrobenzylamine hcl, pubchem12455, acmc-209enh, 4-nitrobenzylamine hcl salt, ksc493k2b, p-nitrobenzylaminehydrochloride, 4-nitrobenzylaminehydrochloride
C1=CC(=CC=C1CN)[N+](=O)[O-].Cl
188.611
188.62

Chemical Identifiers

18600-42-5
188.611
SMIXZZMSWYOQPW-UHFFFAOYSA-N
11252467
C1=CC(=CC=C1CN)[N+](=O)[O-].Cl
C7H9ClN2O2
MFCD00012863
4-nitrobenzylamine hydrochloride, 4-nitrophenyl methanamine hydrochloride, p-nitrobenzylamine hydrochloride, 4-nitrobenzylamine hcl, pubchem12455, acmc-209enh, 4-nitrobenzylamine hcl salt, ksc493k2b, p-nitrobenzylaminehydrochloride, 4-nitrobenzylaminehydrochloride
(4-nitrophenyl)methanamine;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 242-441-3

RTECSNumber : DP6705000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only