missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitrocatechol, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1212214

 View more versions of this product

Catalog No. AAA1212214


Explore available promotions

Description

Description

4-nitrocatechol (4ncat) is a suitable ligand for amperometric determination of Al(III) at environmental concentrations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-nitrocatechol (4ncat) is a suitable ligand for amperometric determination of Al(III) at environmental concentrations.

Solubility
Soluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications

Specifications

4-Nitrocatechol
174°C to 177°C
MFCD00007242
1867508
Soluble in water.
C(=O)[O-].C(=O)[O-].[Ni+2]
148.73
155.11
3316-09-4
C2H2NiO4
25 g
nickel formate, nickel diformate, nickel ii formate, unii-y4yt27n8qn, nickel 2+ diformate, y4yt27n8qn, formic acid, nickel 2+ salt, formic acid, nickel salt, formic acid, nickel 2+ salt 2:1, 64-18-6 parent
HZPNKQREYVVATQ-UHFFFAOYSA-L
nickel(2+);diformate
27506
≥98%

Chemical Identifiers

3316-09-4
MFCD00007242
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 222-009-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only