missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitrophenethyl alcohol, 99%, Thermo Scientific™

Chemical Identifiers

CAS 100-27-6
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.16
InChI Key IKMXRUOZUUKSON-UHFFFAOYSA-N
Synonym 4-nitrophenethyl alcohol, 2-4-nitrophenyl ethanol, 4-nitrobenzeneethanol, 2-p-nitrophenyl ethanol, benzeneethanol, 4-nitro, p-nitrophenethyl alcohol, 2-4-nitrophenyl ethan-1-ol, phenethyl alcohol, p-nitro, ccris 6079, 4-nitrophenethylalcohol
PubChem CID 7494
IUPAC Name 2-(4-nitrophenyl)ethanol
SMILES C1=CC(=CC=C1CCO)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC179610100
SDS View Documents
Thermo Scientific Chemicals
179610100
10g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC179610500
SDS View Documents
Thermo Scientific Chemicals
179610500
50g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

100-27-6
167.16
4-nitrophenethyl alcohol, 2-4-nitrophenyl ethanol, 4-nitrobenzeneethanol, 2-p-nitrophenyl ethanol, benzeneethanol, 4-nitro, p-nitrophenethyl alcohol, 2-4-nitrophenyl ethan-1-ol, phenethyl alcohol, p-nitro, ccris 6079, 4-nitrophenethylalcohol
2-(4-nitrophenyl)ethanol
C8H9NO3
IKMXRUOZUUKSON-UHFFFAOYSA-N
7494
C1=CC(=CC=C1CCO)[N+](=O)[O-]

Specifications

100-27-6
Authentic
C8H9NO3
10g
4-nitrophenethyl alcohol, 2-4-nitrophenyl ethanol, 4-nitrobenzeneethanol, 2-p-nitrophenyl ethanol, benzeneethanol, 4-nitro, p-nitrophenethyl alcohol, 2-4-nitrophenyl ethan-1-ol, phenethyl alcohol, p-nitro, ccris 6079, 4-nitrophenethylalcohol
C1=CC(=CC=C1CCO)[N+](=O)[O-]
167.16
167.16
4-Nitrophenethyl alcohol
148.0°C to 149.0°C (2.0 Torr)
98.5% min. (HPLC)
O2NC6H4CH2CH2OH
06,II,238
IKMXRUOZUUKSON-UHFFFAOYSA-N
2-(4-nitrophenyl)ethanol
7494
99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 202-835-8

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only