Learn More
4-Nitrophenol, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 157050010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorSpecifications
4-Nitrophenol | |
>112°C | |
279°C | |
Authentic | |
Plastic bottle | |
O2NC6H4OH | |
1 kg | |
01,743; 02,297 | |
p-nitrophenol, phenol, 4-nitro, paranitrophenol, niphen, 4-hydroxynitrobenzene, p-hydroxynitrobenzene, phenol, p-nitro, mononitrophenol, paranitrofenol, paranitrofenolo | |
BTJIUGUIPKRLHP-UHFFFAOYSA-N | |
4-nitrophenol | |
980 | |
CHEBI:16836 | |
99% |
100-02-7 | |
Yellow to Brown | |
169°C | |
98.5% min. (GC) | |
C6H5NO3 | |
MFCD00007331 | |
06, 226 | |
15, 6708 | |
Solubility in water: 1.6g/100mL (25 c). Other solubilities: soluble in alcohol,ether,benzene,chloroform and,toluene,pyridine,diethyl ether,also soluble in solns of fixed alkali hydroxides,and carbonates | |
OC1=CC=C(C=C1)[N+]([O-])=O | |
139.11 | |
2.56 mPa.s (121°C) | |
139.11 | |
Crystalline Powder, Crystals or Chunks |
Chemical Identifiers
100-02-7 | |
139.11 | |
BTJIUGUIPKRLHP-UHFFFAOYSA-N | |
980 | |
4-nitrophenol |
C6H5NO3 | |
MFCD00007331 | |
p-nitrophenol, phenol, 4-nitro, paranitrophenol, niphen, 4-hydroxynitrobenzene, p-hydroxynitrobenzene, phenol, p-nitro, mononitrophenol, paranitrofenol, paranitrofenolo | |
CHEBI:16836 | |
OC1=CC=C(C=C1)[N+]([O-])=O |
Safety and Handling
GHS H Statement
May cause damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
Harmful in contact with skin.
Toxic if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
GHS Signal Word: Danger
EINECSNumber : 202-811-7
RUO – Research Use Only