missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitrophenol, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 157050051

 View more versions of this product

Catalog No. AC157050051


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

4-Nitrophenol
>112.0°C
279.0°C
Authentic
Plastic drum
O2NC6H4OH
5 kg
01,743; 02,297
p-nitrophenol, phenol, 4-nitro, paranitrophenol, niphen, 4-hydroxynitrobenzene, p-hydroxynitrobenzene, phenol, p-nitro, mononitrophenol, paranitrofenol, paranitrofenolo
BTJIUGUIPKRLHP-UHFFFAOYSA-N
4-nitrophenol
980
CHEBI:16836
99%
100-02-7
Yellow to Brown
169°C
98.5% min. (GC)
C6H5NO3
MFCD00007331
06, 226
15, 6708
Solubility in water: 1.6g/100mL (25 c). Other solubilities: soluble in alcohol,ether,benzene,chloroform and,toluene,pyridine,diethyl ether,also soluble in solns of fixed alkali hydroxides,and carbonates
OC1=CC=C(C=C1)[N+]([O-])=O
139.11
2.56 mPa.s (121°C)
139.11
Crystalline Powder, Crystals or Chunks

Chemical Identifiers

100-02-7
139.11
BTJIUGUIPKRLHP-UHFFFAOYSA-N
980
4-nitrophenol
C6H5NO3
MFCD00007331
p-nitrophenol, phenol, 4-nitro, paranitrophenol, niphen, 4-hydroxynitrobenzene, p-hydroxynitrobenzene, phenol, p-nitro, mononitrophenol, paranitrofenol, paranitrofenolo
CHEBI:16836
OC1=CC=C(C=C1)[N+]([O-])=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
Harmful in contact with skin.
Toxic if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.

GHS Signal Word: Danger

EINECSNumber : 202-811-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only