missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitrophenyl-alpha-D-glucopyranoside, 98+%, Thermo Scientific Chemicals

$116.88 - $359.75

Chemical Identifiers

CAS 3767-28-0
Molecular Formula C12H15NO8
Molecular Weight (g/mol) 301.251
MDL Number MFCD00064088
InChI Key IFBHRQDFSNCLOZ-ZIQFBCGOSA-N
Synonym 4-nitrophenyl-alpha-d-glucopyranoside, a-d-glucopyranoside, 4-nitrophenyl a-d-glucopyranoside, 4-nitrophenyl alpha-glucoside, pnpalphaglu, pnpalphaglu, p-nitrophenyl alpha-d-glucopyranoside, 4'-nitrophenyl-alpha-d-glucopyranoside, png, p-nitrophenyl, p-nitrophenyl
PubChem CID 92969
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol
SMILES C1=CC(=CC=C1[N+](=O)[O-])OC2C(C(C(C(O2)CO)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1007003
View Documents
Thermo Scientific Chemicals
L1007003
1 g
Each for $116.88
 
AAL1007006
View Documents
Thermo Scientific Chemicals
L1007006
5 g
Each for $359.75
 
Description

Description

4-Nitrophenyl-α-D-glucopyranoside acts as a chromogenic substrate for alfa-D-glucosidase inhibitor. It is also used in the detection of glucansucrases and for yeast alfa-D-glucosidase. Further, it is a substrate for lysosomal alfa-glucosidase and maltase-glucoamylase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophenyl-α-D-glucopyranoside acts as a chromogenic substrate for alfa-D-glucosidase inhibitor. It is also used in the detection of glucansucrases and for yeast alfa-D-glucosidase. Further, it is a substrate for lysosomal alfa-glucosidase and maltase-glucoamylase.

Solubility
Soluble in water, warm ethanol and methanol.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Light sensitive. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

3767-28-0
301.251
IFBHRQDFSNCLOZ-ZIQFBCGOSA-N
92969
C1=CC(=CC=C1[N+](=O)[O-])OC2C(C(C(C(O2)CO)O)O)O
C12H15NO8
MFCD00064088
4-nitrophenyl-alpha-d-glucopyranoside, a-d-glucopyranoside, 4-nitrophenyl a-d-glucopyranoside, 4-nitrophenyl alpha-glucoside, pnpalphaglu, pnpalphaglu, p-nitrophenyl alpha-d-glucopyranoside, 4'-nitrophenyl-alpha-d-glucopyranoside, png, p-nitrophenyl, p-nitrophenyl
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol

Specifications

3767-28-0
C12H15NO8
1 g
4-nitrophenyl-alpha-d-glucopyranoside, a-d-glucopyranoside, 4-nitrophenyl a-d-glucopyranoside, 4-nitrophenyl a-d-glucopyranoside, 4-nitrophenyl alpha-glucoside, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-4-nitrophenoxy oxane-3,4,5-triol, p-nitrophenyl alpha-d-glucopyranoside, p-nitrophenyl alpha-d-glucopyranoside, png, png, p-nitrophenyl
IFBHRQDFSNCLOZ-ZIQFBCGOSA-N
C1=CC(=CC=C1[N+](=O)[O-])OC2C(C(C(C(O2)CO)O)O)O
301.251
301.25
4-Nitrophenyl-alpha-D-glucopyranoside
206°C to 207°C
MFCD00064088
92212
Soluble in water,warm ethanol and methanol.
+218° (c=0.5 in Water)
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol
92969
≥98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 223-189-3

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only