missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Nitrophenyl benzoate, 97%, Thermo Scientific Chemicals
$101.42 - $328.71
Chemical Identifiers
CAS | 959-22-8 |
---|---|
Molecular Formula | C13H9NO4 |
Molecular Weight (g/mol) | 243.218 |
MDL Number | MFCD00135494 |
InChI Key | GMKZBFFLCONHDE-UHFFFAOYSA-N |
Synonym | p-nitrophenyl benzoate, benzoic acid, 4-nitrophenyl ester, benzoic acid, p-nitrophenyl ester, 4-nitrophenyl benzoate, phenol, p-nitro-, benzoate, benzoic acid 4-nitrophenyl ester, benzoic acid,4-nitrophenyl ester |
PubChem CID | 70396 |
IUPAC Name | (4-nitrophenyl) benzoate |
SMILES | C1=CC=C(C=C1)C(=O)OC2=CC=C(C=C2)[N+](=O)[O-] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1602406
|
Thermo Scientific Chemicals
L1602406 |
5 g |
Each for $101.42
|
|
|||||
AAL1602414
|
Thermo Scientific Chemicals
L1602414 |
25 g |
Each for $328.71
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
959-22-8 | |
243.218 | |
GMKZBFFLCONHDE-UHFFFAOYSA-N | |
70396 | |
C1=CC=C(C=C1)C(=O)OC2=CC=C(C=C2)[N+](=O)[O-] |
C13H9NO4 | |
MFCD00135494 | |
p-nitrophenyl benzoate, benzoic acid, 4-nitrophenyl ester, benzoic acid, p-nitrophenyl ester, 4-nitrophenyl benzoate, phenol, p-nitro-, benzoate, benzoic acid 4-nitrophenyl ester, benzoic acid,4-nitrophenyl ester | |
(4-nitrophenyl) benzoate |
Specifications
959-22-8 | |
C13H9NO4 | |
5 g | |
p-nitrophenyl benzoate, benzoic acid, 4-nitrophenyl ester, benzoic acid, p-nitrophenyl ester, 4-nitrophenyl benzoate, phenol, p-nitro-, benzoate, benzoic acid 4-nitrophenyl ester, benzoic acid,4-nitrophenyl ester | |
C1=CC=C(C=C1)C(=O)OC2=CC=C(C=C2)[N+](=O)[O-] | |
243.218 | |
243.22 | |
4-Nitrophenyl benzoate |
142°C to 144°C | |
MFCD00135494 | |
2054709 | |
GMKZBFFLCONHDE-UHFFFAOYSA-N | |
(4-nitrophenyl) benzoate | |
70396 | |
97% |
Safety and Handling
EINECSNumber : 000-000-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only