missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitrophenyl isocyanate, 97%, Thermo Scientific Chemicals

$72.78 - $213.30

Chemical Identifiers

CAS 100-28-7
Molecular Formula C7H4N2O3
Molecular Weight (g/mol) 164.12
MDL Number MFCD00007306
InChI Key GFNKTLQTQSALEJ-UHFFFAOYSA-N
Synonym 4-nitrophenyl isocyanate, p-nitrophenyl isocyanate, benzene, 1-isocyanato-4-nitro, 4-nitrophenylisocyanate, p-nitrophenylisocyanate, isocyanic acid, p-nitrophenyl ester, 1-isocyanato-4-nitro-benzene, isocyanic acid 4-nitrophenyl ester, timtec-bb sbb006628, 4-nitrobenzenisocyanate
PubChem CID 66012
IUPAC Name 1-isocyanato-4-nitrobenzene
SMILES C1=CC(=CC=C1N=C=O)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1119406
View Documents
Thermo Scientific Chemicals
A1119406
5 g
Each for $72.78
 
AAA1119414
View Documents
Thermo Scientific Chemicals
A1119414
25 g
Each for $213.30
 
Description

Description

4-Nitrophenyl isocyanate is used as pharmaceutical intermediates. 4-Nitrophenyl isocyanate was used in the synthesis of 2-(5-methyl-3,4-diphenyl-1H-pyrrole-2-carbonyl)-N-(4-nitrophenyl)hydrazinecarboxamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophenyl isocyanate is used as pharmaceutical intermediates. 4-Nitrophenyl isocyanate was used in the synthesis of 2-(5-methyl-3,4-diphenyl-1H-pyrrole-2-carbonyl)-N-(4-nitrophenyl)hydrazinecarboxamide.

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Store at 2-8°C. Incompatible with water, heat and oxidizing agents.
Specifications

Chemical Identifiers

100-28-7
164.12
GFNKTLQTQSALEJ-UHFFFAOYSA-N
66012
C1=CC(=CC=C1N=C=O)[N+](=O)[O-]
C7H4N2O3
MFCD00007306
4-nitrophenyl isocyanate, p-nitrophenyl isocyanate, benzene, 1-isocyanato-4-nitro, 4-nitrophenylisocyanate, p-nitrophenylisocyanate, isocyanic acid, p-nitrophenyl ester, 1-isocyanato-4-nitro-benzene, isocyanic acid 4-nitrophenyl ester, timtec-bb sbb006628, 4-nitrobenzenisocyanate
1-isocyanato-4-nitrobenzene

Specifications

100-28-7
137°C to 138°C (11 mmHg)
C7H4N2O3
5 g
389516
4-nitrophenyl isocyanate, p-nitrophenyl isocyanate, benzene, 1-isocyanato-4-nitro, 4-nitrophenylisocyanate, p-nitrophenylisocyanate, isocyanic acid, p-nitrophenyl ester, 1-isocyanato-4-nitro-benzene, isocyanic acid 4-nitrophenyl ester, timtec-bb sbb006628, 4-nitrobenzenisocyanate
GFNKTLQTQSALEJ-UHFFFAOYSA-N
1-isocyanato-4-nitrobenzene
66012
97%
56°C to 59°C
>110°C (230°F)
MFCD00007306
UN3335
Moisture sensitive
Insoluble in water.
C1=CC(=CC=C1N=C=O)[N+](=O)[O-]
164.12
164.12
4-Nitrophenyl isocyanate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P332+P313-P342+P311-P362-P501c

H302+H332-H315-H319-H334-H335

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 202-836-3

RTECSNumber : DA3692500

TSCA : Yes

Recommended Storage : Keep cold; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only