missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Nitrophenyl phosphate disodium salt hexahydrate, 5mg tablets, Thermo Scientific Chemicals
Substrate for alkaline and acid phosphatase determinations
$442.68 - $442.68
Chemical Identifiers
CAS | 4264-83-9 |
---|---|
Molecular Formula | C6H4NNa2O6P |
Molecular Weight (g/mol) | 263.05 |
MDL Number | MFCD00007319 |
InChI Key | VIYFPAMJCJLZKD-UHFFFAOYSA-L |
Synonym | pnpp, disodium 4-nitrophenylphosphate, sodium 4-nitrophenyl phosphate, disodium 4-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, disodium salt, disodium p-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, sodium salt 1:2, p-nitrophenyl phosphate, pnpp liquid substrate system |
PubChem CID | 77949 |
IUPAC Name | disodium;(4-nitrophenyl) phosphate |
SMILES | [Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ61401KN
|
Thermo Scientific Chemicals
J61401KN |
100 Pc. |
Each for $442.68
|
|
|||||
Description
4-Nitrophenyl phosphate disodium salt hexahydrate which acts as a chromogenic substrate for the determination of acid and alkaline phosphatases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Nitrophenyl phosphate disodium salt hexahydrate which acts as a chromogenic substrate for the determination of acid and alkaline phosphatases.
Solubility
It is soluble in water.
Notes
The tablets should be stored -20°C
Chemical Identifiers
4264-83-9 | |
263.05 | |
VIYFPAMJCJLZKD-UHFFFAOYSA-L | |
77949 | |
[Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1 |
C6H4NNa2O6P | |
MFCD00007319 | |
pnpp, disodium 4-nitrophenylphosphate, sodium 4-nitrophenyl phosphate, disodium 4-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, disodium salt, disodium p-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, sodium salt 1:2, p-nitrophenyl phosphate, pnpp liquid substrate system | |
disodium;(4-nitrophenyl) phosphate |
Specifications
4264-83-9 | |
C6H4NNa2O6P | |
100 Pc. | |
pnpp, disodium 4-nitrophenylphosphate, sodium 4-nitrophenyl phosphate, disodium 4-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, disodium salt, disodium p-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, sodium salt 1:2, p-nitrophenyl phosphate, pnpp liquid substrate system | |
VIYFPAMJCJLZKD-UHFFFAOYSA-L | |
disodium;(4-nitrophenyl) phosphate | |
77949 | |
Tablets |
Yellow | |
MFCD00007319 | |
5324661 | |
It is soluble in water. | |
[Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1 | |
263.05 | |
371.15 (263.05 Anhydrous) | |
4-Nitrophenyl phosphate disodium salt hexahydrate, 5mg tablets |
Safety and Handling
EINECSNumber : 224-246-5
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only