missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Nitrothiobenzamide, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5214003

 View more versions of this product

Catalog No. AAH5214003


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Nitrothiobenzamide
C7H6N2O2S
1 g
MDBQPBMIZPCKAJ-UHFFFAOYSA-N
4-nitrobenzenecarbothioamide
3000564
97%
26060-30-0
MFCD06150000
4-nitrobenzothioamide, 4-nitro-thiobenzamide, 4-nitrobenzene-1-carbothioamide, 4-nitrothiobenzamide, benzenecarbothioamide,4-nitro, benzenecarbothioamide, 4-nitro, p-nitrothiobenzamide, 4-nitro-benzenecarbothioamide, 4-nitrobenzene-carbothioamide, amino 4-nitrophenyl methane-1-thione
C1=CC(=CC=C1C(=S)N)[N+](=O)[O-]
182.197
182.2

Chemical Identifiers

26060-30-0
182.197
MDBQPBMIZPCKAJ-UHFFFAOYSA-N
3000564
C1=CC(=CC=C1C(=S)N)[N+](=O)[O-]
C7H6N2O2S
MFCD06150000
4-nitrobenzothioamide, 4-nitro-thiobenzamide, 4-nitrobenzene-1-carbothioamide, 4-nitrothiobenzamide, benzenecarbothioamide,4-nitro, benzenecarbothioamide, 4-nitro, p-nitrothiobenzamide, 4-nitro-benzenecarbothioamide, 4-nitrobenzene-carbothioamide, amino 4-nitrophenyl methane-1-thione
4-nitrobenzenecarbothioamide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only