missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Octyne, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04315714

 View more versions of this product

Catalog No. AA4315714


Explore available promotions

Description

Description

4-Octyne, an electron-rich dialkylacetylene, was used in the synthesis of highly substituted 1,3-dienes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Octyne
0.7510 g/mL
20°C (68°F)
C8H14
MFCD00009471
UN3295
Sparingly soluble in water( 29.1mg/L).
CCCC#CCCC
110.2
110.2
Liquid
1942-45-6
131°C to 132°C
Petroleum-like
1.4236
25 g
4-octyne, dipropylacetylene, 1,2-dipropylacetylene, di-n-propylacetylene, acmc-209ey0, 4-octyne 5g, n-c3h7c.$.cc3h7, 3,3?-dihexyloxacarbocyanine iodide
GZTNBKQTTZSQNS-UHFFFAOYSA-N
oct-4-yne
16029
99%

Chemical Identifiers

1942-45-6
110.2
GZTNBKQTTZSQNS-UHFFFAOYSA-N
16029
CCCC#CCCC
C8H14
MFCD00009471
4-octyne, dipropylacetylene, 1,2-dipropylacetylene, di-n-propylacetylene, acmc-209ey0, 4-octyne 5g, n-c3h7c.$.cc3h7, 3,3?-dihexyloxacarbocyanine iodide
oct-4-yne
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c

H226-H304-H315-H319

DOTInformation : Hazard Class: 3; Packaging Group: III

EINECSNumber : 217-730-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only