missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-oxo-4-(2-thienyl)butanoic acid, Thermo Scientific™
Chemical Identifiers
| CAS | 4653-08-1 |
|---|---|
| Molecular Formula | C8H8O3S |
| Molecular Weight (g/mol) | 184.21 |
| InChI Key | ULJMYWHLMLRYSO-UHFFFAOYSA-N |
| Synonym | 3-2-thenoyl propionic acid, 4-oxo-4-thiophen-2-yl butanoic acid, 4-oxo-4-2-thienyl butanoic acid, 4-oxo-4-2-thienyl butyric acid, 3-2-thenoyl propanoic acid, 4-thien-2-yl-4-oxobutyric acid, 4-oxo-4-thien-2-yl butyric acid, 4-thien-2-yl-4-oxobutanoic acid, 4-oxo-4-thien-2-yl butanoic acid, 4-oxo-4-thiophen-2-yl-butyric acid |
| PubChem CID | 78385 |
| ChEBI | CHEBI:64434 |
| IUPAC Name | 4-oxo-4-thiophen-2-ylbutanoic acid |
| SMILES | C1=CSC(=C1)C(=O)CCC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
BTB10644DA
|
Thermo Scientific Chemicals
BTB10644DA |
1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
BTB10644EB
|
Thermo Scientific Chemicals
BTB10644EB |
25g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 4653-08-1 | |
| 184.21 | |
| 3-2-thenoyl propionic acid, 4-oxo-4-thiophen-2-yl butanoic acid, 4-oxo-4-2-thienyl butanoic acid, 4-oxo-4-2-thienyl butyric acid, 3-2-thenoyl propanoic acid, 4-thien-2-yl-4-oxobutyric acid, 4-oxo-4-thien-2-yl butyric acid, 4-thien-2-yl-4-oxobutanoic acid, 4-oxo-4-thien-2-yl butanoic acid, 4-oxo-4-thiophen-2-yl-butyric acid | |
| CHEBI:64434 | |
| C1=CSC(=C1)C(=O)CCC(=O)O |
| C8H8O3S | |
| ULJMYWHLMLRYSO-UHFFFAOYSA-N | |
| 78385 | |
| 4-oxo-4-thiophen-2-ylbutanoic acid |
Specifications
| 4653-08-1 | |
| C8H8O3S | |
| 3-2-thenoyl propionic acid, 4-oxo-4-thiophen-2-yl butanoic acid, 4-oxo-4-2-thienyl butanoic acid, 4-oxo-4-2-thienyl butyric acid, 3-2-thenoyl propanoic acid, 4-thien-2-yl-4-oxobutyric acid, 4-oxo-4-thien-2-yl butyric acid, 4-thien-2-yl-4-oxobutanoic acid, 4-oxo-4-thien-2-yl butanoic acid, 4-oxo-4-thiophen-2-yl-butyric acid | |
| C1=CSC(=C1)C(=O)CCC(=O)O | |
| 184.21 | |
| CHEBI:64434 | |
| 97% |
| 97% | |
| 1g | |
| ULJMYWHLMLRYSO-UHFFFAOYSA-N | |
| 4-oxo-4-thiophen-2-ylbutanoic acid | |
| 78385 | |
| 184.21 | |
| 4-Oxo-4-(2-thienyl)butanoic acid |
Safety and Handling
GHS Signal Word: Warning