missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Pentenal, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1335103

 View more versions of this product

Catalog No. AAL1335103


Explore available promotions

Description

Description

4-Pentenal is used in the preparation of 6-octen-3-one by addition with ethylene using 2,4-pentanedionatobis(ethylene)rhodium(I) as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Pentenal is used in the preparation of 6-octen-3-one by addition with ethylene using 2,4-pentanedionatobis(ethylene)rhodium(I) as a catalyst.

Solubility
Immiscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
Specifications

Specifications

4-Pentenal
0.852
7°C (44°F)
1.417
MFCD00151841
UN1989
Air Sensitive
Immiscible with water.
C=CCCC=O
84.12
84.12
2100-17-6
101°C to 105°C
C5H8O
H2C=CH(CH2)2CHO
1 g
1734369
4-pentenal, 4-penten-1-al, acmc-1cdp8, qumsujwruhpeej-uhfffaoysa
QUMSUJWRUHPEEJ-UHFFFAOYSA-N
pent-4-enal
16418
97%

Chemical Identifiers

2100-17-6
84.12
QUMSUJWRUHPEEJ-UHFFFAOYSA-N
16418
C=CCCC=O
C5H8O
MFCD00151841
4-pentenal, 4-penten-1-al, acmc-1cdp8, qumsujwruhpeej-uhfffaoysa
pent-4-enal
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H302-H312-H332
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P363-P370+P378q-P501c

H225-H302+H312+H332

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALDEHYDES, N.O.S.

EINECSNumber : 218-265-8

RTECSNumber : SB1575000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only