Learn More
4-Pentenoic acid, 98%, Thermo Scientific Chemicals
$73.01 - $504.42
Chemical Identifiers
CAS | 591-80-0 |
---|---|
Molecular Formula | C5H8O2 |
Molecular Weight (g/mol) | 100.12 |
MDL Number | MFCD00004408 |
InChI Key | HVAMZGADVCBITI-UHFFFAOYSA-N |
Synonym | 4-pentenoic acid, allylacetic acid, allyl acetic acid, 4 pa, 3-vinylpropionic acid, delta 4-pentenoic acid, unii-d4s77y29fb, fema no. 2843, .delta.4-pentenoic acid, wln: qv3u1 |
PubChem CID | 61138 |
ChEBI | CHEBI:35936 |
IUPAC Name | pent-4-enoic acid |
SMILES | OC(=O)CCC=C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1399109
|
Thermo Scientific Chemicals
A1399109 |
10 g |
Each for $73.01
|
|
|||||
AAA1399114
|
Thermo Scientific Chemicals
A1399114 |
25 g |
Each for $147.41
|
|
|||||
AAA1399122
|
Thermo Scientific Chemicals
A1399122 |
100 g |
Each for $504.42
|
|
|||||
Description
The sulfonation of 4-pentenoic acid (Allylacetic acid) yields the corresponding γ-lactone. It was used to inhibit fatty acid oxidation in rat heart mitochondria.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThe sulfonation of 4-pentenoic acid (Allylacetic acid) yields the corresponding γ-lactone. It was used to inhibit fatty acid oxidation in rat heart mitochondria.
Solubility
Soluble in alcohol, Slightly soluble in water
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
Chemical Identifiers
591-80-0 | |
100.12 | |
HVAMZGADVCBITI-UHFFFAOYSA-N | |
61138 | |
pent-4-enoic acid |
C5H8O2 | |
MFCD00004408 | |
4-pentenoic acid, allylacetic acid, allyl acetic acid, 4 pa, 3-vinylpropionic acid, delta 4-pentenoic acid, unii-d4s77y29fb, fema no. 2843, .delta.4-pentenoic acid, wln: qv3u1 | |
CHEBI:35936 | |
OC(=O)CCC=C |
Specifications
591-80-0 | |
0.891 | |
89°C (192°F) | |
C5H8O2 | |
MFCD00004408 | |
UN3265 | |
4-pentenoic acid, allylacetic acid, allyl acetic acid, 4 pa, 3-vinylpropionic acid, delta 4-pentenoic acid, unii-d4s77y29fb, fema no. 2843, .delta.4-pentenoic acid, wln: qv3u1 | |
HVAMZGADVCBITI-UHFFFAOYSA-N | |
pent-4-enoic acid | |
61138 | |
100.12 | |
4-Pentenoic acid |
-22°C | |
187°C to 189°C | |
Pungent | |
1.4283 | |
10 g | |
1633696 | |
Soluble in alcohol, Slightly soluble in water | |
OC(=O)CCC=C | |
100.12 | |
CHEBI:35936 | |
98% |
Safety and Handling
GHS H Statement
H314-H318-H302-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Combustible liquid.
P210-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 209-732-7
RTECSNumber : SB2800000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only