missing translation for 'onlineSavingsMsg'
Learn More

4-Pentenoic acid, 99%

CAS: 591-80-0 | C5H8O2 | 100.12 g/mol

$62.19 - $174.89

Chemical Identifiers

CAS 591-80-0
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00004408
InChI Key HVAMZGADVCBITI-UHFFFAOYSA-N
Synonym 4-pentenoic acid, allylacetic acid, allyl acetic acid, 4 pa, 3-vinylpropionic acid, delta 4-pentenoic acid, unii-d4s77y29fb, fema no. 2843, .delta.4-pentenoic acid, wln: qv3u1
PubChem CID 61138
ChEBI CHEBI:35936
IUPAC Name pent-4-enoic acid
SMILES OC(=O)CCC=C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC269020050
SDS View Documents
Thermo Scientific Chemicals
269020050
5 g Glass bottle
Each for $62.19
Only null left
 
AC269020250
SDS View Documents
Thermo Scientific Chemicals
269020250
25 g Glass bottle
Each for $174.89
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

591-80-0
100.12
HVAMZGADVCBITI-UHFFFAOYSA-N
61138
pent-4-enoic acid
C5H8O2
MFCD00004408
4-pentenoic acid, allylacetic acid, allyl acetic acid, 4 pa, 3-vinylpropionic acid, delta 4-pentenoic acid, unii-d4s77y29fb, fema no. 2843, .delta.4-pentenoic acid, wln: qv3u1
CHEBI:35936
OC(=O)CCC=C

Specifications

591-80-0
Colorless to Yellow
83.0°C to 84.0°C (12.0 mmHg)
Authentic
Glass bottle
1.4273 to 1.4303
MFCD00004408
02,425
4-pentenoic acid, allylacetic acid, allyl acetic acid, 4 pa, 3-vinylpropionic acid, delta 4-pentenoic acid, unii-d4s77y29fb, fema no. 2843, .delta.4-pentenoic acid, wln: qv3u1
HVAMZGADVCBITI-UHFFFAOYSA-N
pent-4-enoic acid
61138
100.2
Liquid
-22.5°C
0.9750g/mL
89°C
98.5% min. (GC)
C5H8O2
H2C=CHCH2CH2CO2H
5 g
0.975
Solubility in water: insoluble
OC(=O)CCC=C
100.12
CHEBI:35936
99%
4-Pentenoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 209-732-7

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only