Learn More
4-Pentynoic acid, 95%, Thermo Scientific Chemicals
$114.56 - $366.53
Chemical Identifiers
CAS | 6089-09-4 |
---|---|
Molecular Formula | C5H6O2 |
Molecular Weight (g/mol) | 98.1 |
MDL Number | MFCD00004407 |
InChI Key | MLBYLEUJXUBIJJ-UHFFFAOYSA-M |
Synonym | 4-pentynoic acid, propargylacetic acid, pent-4-yn-1-oic acid, 4-pentynoicacid, pent4-ynoic acid, zlchem 796, pubchem3491, pent-4-yn-oic acid, acmc-1az7d |
PubChem CID | 22464 |
IUPAC Name | pent-4-ynoic acid |
SMILES | C#CCCC(=O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC213730010
|
Thermo Scientific Chemicals
213730010 |
1 g | Glass bottle |
N/A
|
|
|||||
AC213730050
|
Thermo Scientific Chemicals
213730050 |
5 g | Glass bottle |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6089-09-4 | |
98.1 | |
MLBYLEUJXUBIJJ-UHFFFAOYSA-M | |
22464 | |
C#CCCC(=O)O |
C5H6O2 | |
MFCD00004407 | |
4-pentynoic acid, propargylacetic acid, pent-4-yn-1-oic acid, 4-pentynoicacid, pent4-ynoic acid, zlchem 796, pubchem3491, pent-4-yn-oic acid, acmc-1az7d | |
pent-4-ynoic acid |
Specifications
6089-09-4 | |
100.0 | |
White to Beige | |
75°C | |
94% min. (GC) | |
C5H6O2 | |
MFCD00004407 | |
02, 481 | |
Solubility in water: soluble. Other solubilities: soluble in ethanol,acetone and,methyl-t.-buthylether | |
C#CCCC(=O)O | |
98.1 | |
98.1 | |
Crystalline Powder or Flakes |
97.5 | |
54.0°C to 58.0°C | |
110.0°C (30.0 mmHg) | |
Authentic | |
Glass bottle | |
HC≡CCH2CH2CO2H | |
1 g | |
4-pentynoic acid, propargylacetic acid, pent-4-yn-1-oic acid, 4-pentynoicacid, pent4-ynoic acid, zlchem 796, pubchem3491, pent-4-yn-oic acid, acmc-1az7d | |
MLBYLEUJXUBIJJ-UHFFFAOYSA-M | |
pent-4-ynoic acid | |
22464 | |
95% | |
4-Pentynoic acid, 95% |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for severa
GHS Signal Word: Danger
EINECSNumber : 228-028-0
RTECSNumber : SC4751000
RUO – Research Use Only