missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(4-Phenoxyphenyl)methylamine hydrochloride, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 169944-04-1 |
---|---|
Molecular Formula | C13H14ClNO |
Molecular Weight (g/mol) | 235.711 |
InChI Key | VHCSCKHIGGFTHN-UHFFFAOYSA-N |
Synonym | 4-phenoxyphenyl methanamine hydrochloride, 4-phenoxyphenyl methylamine hydrochloride, 4-phenoxybenzylamine hydrochloride, benzenemethanamine, 4-phenoxy-, hydrochloride, 1-4-phenoxyphenyl methanamine hydrochloride, 4-phenoxyphenyl methylamine, chloride, 1-4-phenoxyphenyl methanamine-hydrogen chloride 1/1 |
PubChem CID | 22293026 |
IUPAC Name | (4-phenoxyphenyl)methanamine;hydrochloride |
SMILES | C1=CC=C(C=C1)OC2=CC=C(C=C2)CN.Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
CC53763DA
|
Thermo Scientific Chemicals
CC53763DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC53763DE
|
Thermo Scientific Chemicals
CC53763DE |
5g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
169944-04-1 | |
235.711 | |
4-phenoxyphenyl methanamine hydrochloride, 4-phenoxyphenyl methylamine hydrochloride, 4-phenoxybenzylamine hydrochloride, benzenemethanamine, 4-phenoxy-, hydrochloride, 1-4-phenoxyphenyl methanamine hydrochloride, 4-phenoxyphenyl methylamine, chloride, 1-4-phenoxyphenyl methanamine-hydrogen chloride 1/1 | |
(4-phenoxyphenyl)methanamine;hydrochloride |
C13H14ClNO | |
VHCSCKHIGGFTHN-UHFFFAOYSA-N | |
22293026 | |
C1=CC=C(C=C1)OC2=CC=C(C=C2)CN.Cl |
Specifications
169944-04-1 | |
Amber Glass Bottle | |
1g | |
VHCSCKHIGGFTHN-UHFFFAOYSA-N | |
(4-phenoxyphenyl)methanamine;hydrochloride | |
22293026 | |
97% |
97% | |
C13H14ClNO | |
4-phenoxyphenyl methanamine hydrochloride, 4-phenoxyphenyl methylamine hydrochloride, 4-phenoxybenzylamine hydrochloride, benzenemethanamine, 4-phenoxy-, hydrochloride, 1-4-phenoxyphenyl methanamine hydrochloride, 4-phenoxyphenyl methylamine, chloride, 1-4-phenoxyphenyl methanamine-hydrogen chloride 1/1 | |
C1=CC=C(C=C1)OC2=CC=C(C=C2)CN.Cl | |
235.711 | |
235.71 | |
(4-Phenoxyphenyl)methylamine hydrochloride |
Safety and Handling
GHS Signal Word: Warning