missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Phenyl-1-butyne, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 449770050

 View more versions of this product

Catalog No. AC449770050



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Phenyl-1-butyne
0.9260g/mL
66°C
C10H10
0.926
Solubility in water: insoluble
C#CCCC1=CC=CC=C1
130.19
130.19
16520-62-0
190.0°C
96% min. (GC)
5 g
4-phenyl-1-butyne, 3-butynylbenzene, but-3-yn-1-ylbenzene, benzene, 3-butynyl, 2-phenylethylacetylene, ccris 7100, phenethylacetylene, 4-phenyl-1-butyn, 3-butynylbenzene #, 4-phenylbut-1-yne
QDEOKXOYHYUKMS-UHFFFAOYSA-N
but-3-ynylbenzene
123360
97%

Chemical Identifiers

16520-62-0
130.19
4-phenyl-1-butyne, 3-butynylbenzene, but-3-yn-1-ylbenzene, benzene, 3-butynyl, 2-phenylethylacetylene, ccris 7100, phenethylacetylene, 4-phenyl-1-butyn, 3-butynylbenzene #, 4-phenylbut-1-yne
but-3-ynylbenzene
C10H10
QDEOKXOYHYUKMS-UHFFFAOYSA-N
123360
C#CCCC1=CC=CC=C1
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only