missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyl-2-butanone, 98%

CAS: 2550-26-7 | C10H12O | 148.205 g/mol

$117.92 - $285.99

Chemical Identifiers

CAS 2550-26-7
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00008790
InChI Key AKGGYBADQZYZPD-UHFFFAOYSA-N
Synonym benzylacetone, 4-phenyl-2-butanone, 2-butanone, 4-phenyl, 4-penylbutan-2-one, 1-phenyl-3-butanone, methyl phenethyl ketone, benzyl acetone, methyl 2-phenylethyl ketone, phenethyl methyl ketone, methyl phenylethyl ketone
PubChem CID 17355
IUPAC Name 4-phenylbutan-2-one
SMILES CC(=O)CCC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2277730
View Documents
Thermo Scientific Chemicals
B2277730
250 g N/A
Only null left
 
AAB227770B
View Documents
Thermo Scientific Chemicals
B227770B
1000 g N/A
Only null left
 
Description

Description

4-Phenyl-2-butanone be used as an attractant for melon flies and as an odorant for soap. It is also used in the preparation of 4-oxocyclohexanecarbaldehyde derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Phenyl-2-butanone be used as an attractant for melon flies and as an odorant for soap. It is also used in the preparation of 4-oxocyclohexanecarbaldehyde derivatives.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

2550-26-7
148.205
AKGGYBADQZYZPD-UHFFFAOYSA-N
17355
CC(=O)CCC1=CC=CC=C1
C10H12O
MFCD00008790
benzylacetone, 4-phenyl-2-butanone, 2-butanone, 4-phenyl, 4-penylbutan-2-one, 1-phenyl-3-butanone, methyl phenethyl ketone, benzyl acetone, methyl 2-phenylethyl ketone, phenethyl methyl ketone, methyl phenylethyl ketone
4-phenylbutan-2-one

Specifications

2550-26-7
235°C to 237°C
C10H12O
MFCD00008790
1907123
Not miscible in water.
CC(=O)CCC1=CC=CC=C1
148.205
148.21
4-Phenyl-2-butanone
0.988
105°C (221°F)
1.5122
250 g
benzylacetone, 4-phenyl-2-butanone, 2-butanone, 4-phenyl, 4-penylbutan-2-one, 1-phenyl-3-butanone, methyl phenethyl ketone, benzyl acetone, methyl 2-phenylethyl ketone, phenethyl methyl ketone, methyl phenylethyl ketone
AKGGYBADQZYZPD-UHFFFAOYSA-N
4-phenylbutan-2-one
17355
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 219-847-4

RTECSNumber : EL9600000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only