missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Phenylazo)benzoic Acid 98.0+%, TCI America™
$72.70 - $238.60
Chemical Identifiers
| CAS | 1562-93-2 |
|---|---|
| Molecular Formula | C13H10N2O2 |
| Molecular Weight (g/mol) | 226.235 |
| MDL Number | MFCD00020348 |
| InChI Key | CSPTZWQFHBVOLO-UHFFFAOYSA-N |
| Synonym | Azobenzene-4-carboxylic Acid |
| PubChem CID | 15276 |
| IUPAC Name | 4-phenyldiazenylbenzoic acid |
| SMILES | C1=CC=C(C=C1)N=NC2=CC=C(C=C2)C(=O)O |
Chemical Identifiers
| 1562-93-2 | |
| 226.235 | |
| CSPTZWQFHBVOLO-UHFFFAOYSA-N | |
| 15276 | |
| C1=CC=C(C=C1)N=NC2=CC=C(C=C2)C(=O)O |
| C13H10N2O2 | |
| MFCD00020348 | |
| Azobenzene-4-carboxylic Acid | |
| 4-phenyldiazenylbenzoic acid |
Specifications
| 1562-93-2 | |
| C13H10N2O2 | |
| 1 g | |
| CSPTZWQFHBVOLO-UHFFFAOYSA-N | |
| 4-phenyldiazenylbenzoic acid | |
| 15276 | |
| ≥98.0% (T) | |
| 4-(Phenylazo)benzoic Acid |
| 248°C | |
| MFCD00020348 | |
| Azobenzene-4-carboxylic Acid | |
| C1=CC=C(C=C1)N=NC2=CC=C(C=C2)C(=O)O | |
| 226.235 | |
| 226.24 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes