missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylbutyric acid, 99%

CAS: 1821-12-1 | C10H12O2 | 164.2 g/mol

$128.36 - $128.36

Chemical Identifiers

CAS 1821-12-1
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.2
InChI Key OBKXEAXTFZPCHS-UHFFFAOYSA-N
Synonym 4-phenylbutyric acid, benzenebutanoic acid, benzenebutyric acid, phenylbutyrate, gamma-phenylbutyric acid, omega-phenylbutanoic acid, phenylbutyric acid, 4-phenylbutyrate, butyric acid, 4-phenyl, 4-phenyl-n-butyric acid
PubChem CID 4775
ChEBI CHEBI:41500
IUPAC Name 4-phenylbutanoic acid
SMILES C1=CC=C(C=C1)CCCC(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC130380250
SDS View Documents
Thermo Scientific Chemicals
130380250
25 g Glass bottle
Each for $128.36
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1821-12-1
164.2
4-phenylbutyric acid, benzenebutanoic acid, benzenebutyric acid, phenylbutyrate, gamma-phenylbutyric acid, omega-phenylbutanoic acid, phenylbutyric acid, 4-phenylbutyrate, butyric acid, 4-phenyl, 4-phenyl-n-butyric acid
CHEBI:41500
C1=CC=C(C=C1)CCCC(=O)O
C10H12O2
OBKXEAXTFZPCHS-UHFFFAOYSA-N
4775
4-phenylbutanoic acid

Specifications

1821-12-1
White to Yellow
>110°C
99%
C10H12O2
25 g
4-phenylbutyric acid, benzenebutanoic acid, benzenebutyric acid, phenylbutyrate, gamma-phenylbutyric acid, omega-phenylbutanoic acid, phenylbutyric acid, 4-phenylbutyrate, butyric acid, 4-phenyl, 4-phenyl-n-butyric acid
OBKXEAXTFZPCHS-UHFFFAOYSA-N
4-phenylbutanoic acid
4775
164.2
Crystalline Powder
49.0°C to 52.0°C
165.0°C (10.0 mmHg)
Authentic
Glass bottle
C6H5(CH2)3CO2H
09,539
Solubility in water: 5.3g/L water (40 degr. c)
C1=CC=C(C=C1)CCCC(=O)O
164.2
CHEBI:41500
99%
4-Phenylbutyric acid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 217-341-8

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only