Learn More
4-Phthalimido-2-butyne, 97%, Thermo Scientific™
Identifiants chimiques
CAS | 113439-83-1 |
---|---|
Molecular Formula | C12H9NO2 |
Molecular Weight (g/mol) | 199.21 |
MDL Number | MFCD00160830 |
InChI Key | COPWPECHFWIYSP-UHFFFAOYSA-N |
Synonym | n-2-butynyl phthalimide, 2-but-2-yn-1-yl isoindoline-1,3-dione, 2-but-2-yn-1-yl isoindole-1,3-dione, acmc-1bscb, 1-phthalimido-2-butyne, 4-phthalimido-2-butyne, 2-2-butynyl isoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-2-butyn-1-yl |
PubChem CID | 8063352 |
IUPAC Name | 2-(but-2-yn-1-yl)-2,3-dihydro-1H-isoindole-1,3-dione |
SMILES | CC#CCN1C(=O)C2=CC=CC=C2C1=O |
Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
---|---|---|---|---|---|---|---|---|---|
Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
AC382150010
|
Thermo Scientific Chemicals
382150010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
Identifiants chimiques
113439-83-1 | |
199.21 | |
COPWPECHFWIYSP-UHFFFAOYSA-N | |
8063352 | |
CC#CCN1C(=O)C2=CC=CC=C2C1=O |
C12H9NO2 | |
MFCD00160830 | |
n-2-butynyl phthalimide, 2-but-2-yn-1-yl isoindoline-1,3-dione, 2-but-2-yn-1-yl isoindole-1,3-dione, acmc-1bscb, 1-phthalimido-2-butyne, 4-phthalimido-2-butyne, 2-2-butynyl isoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-2-butyn-1-yl | |
2-(but-2-yn-1-yl)-2,3-dihydro-1H-isoindole-1,3-dione |
Spécifications
113439-83-1 | |
100.0 | |
96% min. (GC) | |
C12H9NO2 | |
1g | |
COPWPECHFWIYSP-UHFFFAOYSA-N | |
2-(but-2-yn-1-yl)-2,3-dihydro-1H-isoindole-1,3-dione | |
8063352 | |
97% |
96.0 | |
Authentic | |
Glass bottle | |
MFCD00160830 | |
n-2-butynyl phthalimide, 2-but-2-yn-1-yl isoindoline-1,3-dione, 2-but-2-yn-1-yl isoindole-1,3-dione, acmc-1bscb, 1-phthalimido-2-butyne, 4-phthalimido-2-butyne, 2-2-butynyl isoindoline-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-2-butyn-1-yl | |
CC#CCN1C(=O)C2=CC=CC=C2C1=O | |
199.21 | |
199.21 | |
4-Phthalimido-2-butyne |
Sécurité et manipulation
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO â Research Use Only