missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Propargyloxy)benzaldehyde 98.0+%, TCI America™
$134.21 - $536.84
Chemical Identifiers
| CAS | 5651-86-5 |
|---|---|
| Molecular Formula | C10H8O2 |
| Molecular Weight (g/mol) | 160.17 |
| MDL Number | MFCD01088150 |
| InChI Key | GSSBOYWRKTVVQX-UHFFFAOYSA-N |
| Synonym | 4-(2-Propynyloxy)benzaldehyde |
| PubChem CID | 1897533 |
| IUPAC Name | 4-(prop-2-yn-1-yloxy)benzaldehyde |
| SMILES | O=CC1=CC=C(OCC#C)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P2339200MG
|
TCI America
P2339200MG |
200 mg |
Each for $134.21
|
|
|||||
|
P23391G
|
TCI America
P23391G |
1 g |
Each for $536.84
|
|
|||||
Chemical Identifiers
| 5651-86-5 | |
| 160.17 | |
| GSSBOYWRKTVVQX-UHFFFAOYSA-N | |
| 1897533 | |
| O=CC1=CC=C(OCC#C)C=C1 |
| C10H8O2 | |
| MFCD01088150 | |
| 4-(2-Propynyloxy)benzaldehyde | |
| 4-(prop-2-yn-1-yloxy)benzaldehyde |
Specifications
| 5651-86-5 | |
| C10H8O2 | |
| 200 mg | |
| GSSBOYWRKTVVQX-UHFFFAOYSA-N | |
| 4-(prop-2-yn-1-yloxy)benzaldehyde | |
| 1897533 | |
| ≥98.0% (GC) | |
| 4-(Propargyloxy)benzaldehyde |
| 82°C | |
| MFCD01088150 | |
| 4-(2-Propynyloxy)benzaldehyde | |
| O=CC1=CC=C(OCC#C)C=C1 | |
| 160.17 | |
| 160.17 | |
| Crystalline Powder |
Safety and Handling
TSCA : No