missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Propoxyacetophenone 98.0+%, TCI America™
$124.89 - $378.40
Chemical Identifiers
| CAS | 5736-86-7 |
|---|---|
| Molecular Formula | C11H14O2 |
| Molecular Weight (g/mol) | 178.231 |
| MDL Number | MFCD01922144 |
| InChI Key | RTYYKCQJSTZADZ-UHFFFAOYSA-N |
| PubChem CID | 260132 |
| IUPAC Name | 1-(4-propoxyphenyl)ethanone |
| SMILES | CCCOC1=CC=C(C=C1)C(=O)C |
Chemical Identifiers
| 5736-86-7 | |
| 178.231 | |
| RTYYKCQJSTZADZ-UHFFFAOYSA-N | |
| 1-(4-propoxyphenyl)ethanone |
| C11H14O2 | |
| MFCD01922144 | |
| 260132 | |
| CCCOC1=CC=C(C=C1)C(=O)C |
Specifications
| 5736-86-7 | |
| White | |
| C11H14O2 | |
| 1 g | |
| CCCOC1=CC=C(C=C1)C(=O)C | |
| 178.231 | |
| 178.23 | |
| Crystalline Lumps |
| 21°C | |
| 111°C | |
| MFCD01922144 | |
| RTYYKCQJSTZADZ-UHFFFAOYSA-N | |
| 1-(4-propoxyphenyl)ethanone | |
| 260132 | |
| ≥98.0% (GC) | |
| 4′-Propoxyacetophenone |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator