missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Propylbenzoic Acid 99.0+%, TCI America™
$137.94 - $723.25
Chemical Identifiers
| CAS | 2438-05-3 |
|---|---|
| Molecular Formula | C10H11O2 |
| Molecular Weight (g/mol) | 163.20 |
| MDL Number | MFCD00013996 |
| InChI Key | ATZHGRNFEFVDDJ-UHFFFAOYSA-M |
| Synonym | 4-n-propylbenzoic acid, benzoic acid, 4-propyl, 4-propyl-benzoic acid, p-n-propyl benzoic acid, p-propylbenzoic acid, unii-3p1v85e45o, 4-propylbenzoicacid, p-propyl benzoic acid, 4-propylbenzolic acid, pubchem1348 |
| PubChem CID | 137601 |
| IUPAC Name | 4-propylbenzoic acid |
| SMILES | CCCC1=CC=C(C=C1)C(=O)O |
Chemical Identifiers
| 2438-05-3 | |
| 163.20 | |
| ATZHGRNFEFVDDJ-UHFFFAOYSA-M | |
| 137601 | |
| CCCC1=CC=C(C=C1)C(=O)O |
| C10H11O2 | |
| MFCD00013996 | |
| 4-n-propylbenzoic acid, benzoic acid, 4-propyl, 4-propyl-benzoic acid, p-n-propyl benzoic acid, p-propylbenzoic acid, unii-3p1v85e45o, 4-propylbenzoicacid, p-propyl benzoic acid, 4-propylbenzolic acid, pubchem1348 | |
| 4-propylbenzoic acid |
Specifications
| 2438-05-3 | |
| White | |
| MFCD00013996 | |
| 4-n-propylbenzoic acid, benzoic acid, 4-propyl, 4-propyl-benzoic acid, p-n-propyl benzoic acid, p-propylbenzoic acid, unii-3p1v85e45o, 4-propylbenzoicacid, p-propyl benzoic acid, 4-propylbenzolic acid, pubchem1348 | |
| CCCC1=CC=C(C=C1)C(=O)O | |
| 163.20 | |
| 164.20 | |
| Crystalline Powder |
| 142°C | |
| C10H11O2 | |
| 25 g | |
| ATZHGRNFEFVDDJ-UHFFFAOYSA-M | |
| 4-propylbenzoic acid | |
| 137601 | |
| ≥99.0% (T) | |
| 4-Propylbenzoic Acid |
Safety and Handling
TSCA : No