Learn More
4-(Pyrid-2-yloxy)benzoic acid, ≥97%, Thermo Scientific™
$171.50 - $475.88
Chemical Identifiers
CAS | 51363-00-9 |
---|---|
Molecular Formula | C12H9NO3 |
Molecular Weight (g/mol) | 215.21 |
MDL Number | MFCD08435897 |
InChI Key | GKSKQZLHPWBLJL-UHFFFAOYSA-N |
Synonym | 4-pyrid-2-yloxy benzoic acid, 4-pyridin-2-yloxy benzoic acid, 4-2-pyridyloxy benzoic acid, 4-pyridin-2-yloxy-benzoic acid, 4-pyridin-2-yl oxy benzoic acid |
PubChem CID | 22262052 |
IUPAC Name | 4-pyridin-2-yloxybenzoic acid |
SMILES | OC(=O)C1=CC=C(OC2=CC=CC=N2)C=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
CC48901CB
|
Thermo Scientific Chemicals
CC48901CB |
250 mg | Amber Glass Bottle |
Each for $171.50
|
|
||||
CC48901DA
|
Thermo Scientific Chemicals
CC48901DA |
1 g | Amber Glass Bottle |
Each for $475.88
|
|
||||
Chemical Identifiers
51363-00-9 | |
215.21 | |
GKSKQZLHPWBLJL-UHFFFAOYSA-N | |
22262052 | |
OC(=O)C1=CC=C(OC2=CC=CC=N2)C=C1 |
C12H9NO3 | |
MFCD08435897 | |
4-pyrid-2-yloxy benzoic acid, 4-pyridin-2-yloxy benzoic acid, 4-2-pyridyloxy benzoic acid, 4-pyridin-2-yloxy-benzoic acid, 4-pyridin-2-yl oxy benzoic acid | |
4-pyridin-2-yloxybenzoic acid |
Specifications
51363-00-9 | |
100.0 | |
Amber Glass Bottle | |
MFCD08435897 | |
4-pyrid-2-yloxy benzoic acid, 4-pyridin-2-yloxy benzoic acid, 4-2-pyridyloxy benzoic acid, 4-pyridin-2-yloxy-benzoic acid, 4-pyridin-2-yl oxy benzoic acid | |
OC(=O)C1=CC=C(OC2=CC=CC=N2)C=C1 | |
215.21 | |
215.21 | |
4-(Pyrid-2-yloxy)benzoic acid |
96.5 | |
97% | |
C12H9NO3 | |
250 mg | |
GKSKQZLHPWBLJL-UHFFFAOYSA-N | |
4-pyridin-2-yloxybenzoic acid | |
22262052 | |
97% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.