missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Sulfobenzoic acid monopotassium salt, 95%, Thermo Scientific Chemicals

$96.25 - $345.15

Chemical Identifiers

CAS 5399-63-3
Molecular Formula C7H5KO5S
Molecular Weight (g/mol) 240.27
MDL Number MFCD00007509
InChI Key PXRJBUPXKDXDLG-UHFFFAOYSA-M
Synonym benzoic acid, 4-sulfo-, monopotassium salt, 4-sulfobenzoic acid monopotassium salt, 4-sulphobenzoic acid monopotassium salt, 4-sulfobenzoicacidmonopotassiumsalt, potassium hydrogen 4-sulphonatobenzoate, p-sulphobenzoic acid, monopotassium salt, potassium 4-sulphobenzoate, benzoic acid, 4-sulfo-, potassium salt 1:1, potassium 4-sulfobenzoate, p-potassiooxysulfonyl benzoic acid
PubChem CID 23670833
IUPAC Name potassium;4-sulfobenzoate
SMILES C1=CC(=CC=C1C(=O)[O-])S(=O)(=O)O.[K+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
AAA1943218
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
A1943218
50 g
Each for $96.25
 
AAA1943230
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
A1943230
250 g
Each for $345.15
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5399-63-3
240.27
PXRJBUPXKDXDLG-UHFFFAOYSA-M
23670833
C1=CC(=CC=C1C(=O)[O-])S(=O)(=O)O.[K+]
C7H5KO5S
MFCD00007509
benzoic acid, 4-sulfo-, monopotassium salt, 4-sulfobenzoic acid monopotassium salt, 4-sulphobenzoic acid monopotassium salt, 4-sulfobenzoicacidmonopotassiumsalt, potassium hydrogen 4-sulphonatobenzoate, p-sulphobenzoic acid, monopotassium salt, potassium 4-sulphobenzoate, benzoic acid, 4-sulfo-, potassium salt 1:1, potassium 4-sulfobenzoate, p-potassiooxysulfonyl benzoic acid
potassium;4-sulfobenzoate

Specifications

5399-63-3
MFCD00007509
Hygroscopic
PXRJBUPXKDXDLG-UHFFFAOYSA-M
potassium;4-sulfobenzoate
23670833
95%
C7H5KO5S
50 g
benzoic acid, 4-sulfo-, monopotassium salt, 4-sulfobenzoic acid monopotassium salt, 4-sulphobenzoic acid monopotassium salt, 4-sulfobenzoicacidmonopotassiumsalt, potassium hydrogen 4-sulphonatobenzoate, p-sulphobenzoic acid, monopotassium salt, potassium 4-sulphobenzoate, benzoic acid, 4-sulfo-, potassium salt 1:1, potassium 4-sulfobenzoate, p-potassiooxysulfonyl benzoic acid
C1=CC(=CC=C1C(=O)[O-])S(=O)(=O)O.[K+]
240.27
240.28
4-Sulfobenzoic acid monopotassium salt
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 226-435-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only