Learn More
4-tert-Amylphenol, 99%
CAS: 80-46-6 | C11H16O | 164.25 g/mol
$50.49 - $281.76
Chemical Identifiers
| CAS | 80-46-6 |
|---|---|
| Molecular Formula | C11H16O |
| Molecular Weight (g/mol) | 164.25 |
| MDL Number | MFCD00002369 |
| InChI Key | NRZWYNLTFLDQQX-UHFFFAOYSA-N |
| Synonym | 4-tert-amylphenol, 4-tert-pentylphenol, p-tert-amylphenol, p-tert-pentylphenol, 4-t-amylphenol, 4-1,1-dimethylpropyl phenol, amilphenol, amilfenol, pentaphen, tert-amylphenol |
| PubChem CID | 6643 |
| ChEBI | CHEBI:35096 |
| IUPAC Name | 4-(2-methylbutan-2-yl)phenol |
| SMILES | CCC(C)(C)C1=CC=C(C=C1)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC167330050
|
Thermo Scientific Chemicals
167330050 |
5 g |
Each for $50.49
|
|
|||||
|
AC167331000
|
Thermo Scientific Chemicals
167331000 |
100 g |
Each for $80.22
|
|
|||||
|
AC167332500
|
Thermo Scientific Chemicals
167332500 |
250 g |
Each for $133.89
|
|
|||||
|
AC167335000
|
Thermo Scientific Chemicals
167335000 |
500 g |
Each for $281.76
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 80-46-6 | |
| 164.25 | |
| NRZWYNLTFLDQQX-UHFFFAOYSA-N | |
| 6643 | |
| 4-(2-methylbutan-2-yl)phenol |
| C11H16O | |
| MFCD00002369 | |
| 4-tert-amylphenol, 4-tert-pentylphenol, p-tert-amylphenol, p-tert-pentylphenol, 4-t-amylphenol, 4-1,1-dimethylpropyl phenol, amilphenol, amilfenol, pentaphen, tert-amylphenol | |
| CHEBI:35096 | |
| CCC(C)(C)C1=CC=C(C=C1)O |
Specifications
| 80-46-6 | |
| 100.0 | |
| White to Yellow | |
| 111°C | |
| 98.5% min. (GC) | |
| C11H16O | |
| MFCD00002369 | |
| 06, 548 | |
| 4-tert-amylphenol, 4-tert-pentylphenol, p-tert-amylphenol, p-tert-pentylphenol, 4-t-amylphenol, 4-1,1-dimethylpropyl phenol, amilphenol, amilfenol, pentaphen, tert-amylphenol | |
| NRZWYNLTFLDQQX-UHFFFAOYSA-N | |
| 4-(2-methylbutan-2-yl)phenol | |
| 6643 | |
| CHEBI:35096 | |
| 99% | |
| 4-tert-Amylphenol, 99% |
| 98.5 | |
| >91.0°C | |
| 255.0°C | |
| Authentic | |
| Glass bottle | |
| C2H5C(CH3)2C6H4OH | |
| 5 g | |
| 15, 7255 | |
| Solubility in water: insoluble. Other solubilities: soluble in alcohol, ether, benzene and chloroform | |
| CCC(C)(C)C1=CC=C(C=C1)O | |
| 164.25 | |
| 2.9 mPa.s (100°C) | |
| 164.25 | |
| Briquettes or Flakes to Coarse Powder |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several m
GHS Signal Word: Danger
EINECSNumber : 201-280-9
RTECSNumber : SM6825000
TSCA : TSCA
RUO – Research Use Only