missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(tert-Butoxycarbonylamino)benzoic Acid 98.0+%, TCI America™

$65.17 - $207.38

Chemical Identifiers

CAS 66493-39-8
Molecular Formula C12H15NO4
Molecular Weight (g/mol) 237.255
MDL Number MFCD00037428
InChI Key ZJDBQMWMDZEONW-UHFFFAOYSA-N
Synonym boc-4-abz-oh, 4-boc-amino benzoic acid, 4-tert-butoxycarbonyl amino benzoic acid, 4-tert-butoxycarbonylamino-benzoic acid, n-boc-4-aminobenzoic acid, 4-tert-butoxycarbonylaminobenzoic acid, 4-tert-butoxycarbonylamino benzoic acid, boc-p-aminobenzoic acid, 4-boc-amino-benzoic acid
PubChem CID 2755931
IUPAC Name 4-[(2-methylpropan-2-yl)oxycarbonylamino]benzoic acid
SMILES CC(C)(C)OC(=O)NC1=CC=C(C=C1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B38985G
View Documents
TCI America
B38985G
5 g
Each for $65.17
 
B389825G
View Documents
TCI America
B389825G
25 g
Each for $207.38
 
Description

Description

Specifications

Chemical Identifiers

66493-39-8
237.255
ZJDBQMWMDZEONW-UHFFFAOYSA-N
2755931
CC(C)(C)OC(=O)NC1=CC=C(C=C1)C(=O)O
C12H15NO4
MFCD00037428
boc-4-abz-oh, 4-boc-amino benzoic acid, 4-tert-butoxycarbonyl amino benzoic acid, 4-tert-butoxycarbonylamino-benzoic acid, n-boc-4-aminobenzoic acid, 4-tert-butoxycarbonylaminobenzoic acid, 4-tert-butoxycarbonylamino benzoic acid, boc-p-aminobenzoic acid, 4-boc-amino-benzoic acid
4-[(2-methylpropan-2-yl)oxycarbonylamino]benzoic acid

Specifications

66493-39-8
White
MFCD00037428
boc-4-abz-oh, 4-boc-amino benzoic acid, 4-tert-butoxycarbonyl amino benzoic acid, 4-tert-butoxycarbonylamino-benzoic acid, n-boc-4-aminobenzoic acid, 4-tert-butoxycarbonylaminobenzoic acid, 4-tert-butoxycarbonylamino benzoic acid, boc-p-aminobenzoic acid, 4-boc-amino-benzoic acid
CC(C)(C)OC(=O)NC1=CC=C(C=C1)C(=O)O
237.255
237.26
Crystalline Powder
196°C
C12H15NO4
5 g
ZJDBQMWMDZEONW-UHFFFAOYSA-N
4-[(2-methylpropan-2-yl)oxycarbonylamino]benzoic acid
2755931
≥98.0% (HPLC,T)
4-(tert-Butoxycarbonylamino)benzoic Acid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'