missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-[(tert-Butoxycarbonylamino)methyl]benzoic Acid 98.0+%, TCI America™
$71.33 - $247.19
Chemical Identifiers
CAS | 33233-67-9 |
---|---|
Molecular Formula | C13H16NO4 |
Molecular Weight (g/mol) | 250.28 |
MDL Number | MFCD00228182 |
InChI Key | LNKHBRDWRIIROP-UHFFFAOYSA-M |
Synonym | 4-tert-butoxycarbonylamino methyl benzoic acid, 4-tert-butoxycarbonyl amino methyl benzoic acid, 4-tert-butoxy carbonyl amino methyl benzoic acid, boc-4-aminomethyl-benzoic acid, 4-boc-aminomethyl-benzoic acid, 4-tert-butoxycarbonylaminomethyl benzoic acid, 4-n-tert-butyloxycarbonyl amino methyl benzoic acid, benzoic acid, 4-1,1-dimethylethoxy carbonyl amino methyl |
PubChem CID | 853605 |
IUPAC Name | 4-({[(tert-butoxy)carbonyl]amino}methyl)benzoate |
SMILES | CC(C)(C)OC(=O)NCC1=CC=C(C=C1)C([O-])=O |
Chemical Identifiers
33233-67-9 | |
250.28 | |
LNKHBRDWRIIROP-UHFFFAOYSA-M | |
853605 | |
CC(C)(C)OC(=O)NCC1=CC=C(C=C1)C([O-])=O |
C13H16NO4 | |
MFCD00228182 | |
4-tert-butoxycarbonylamino methyl benzoic acid, 4-tert-butoxycarbonyl amino methyl benzoic acid, 4-tert-butoxy carbonyl amino methyl benzoic acid, boc-4-aminomethyl-benzoic acid, 4-boc-aminomethyl-benzoic acid, 4-tert-butoxycarbonylaminomethyl benzoic acid, 4-n-tert-butyloxycarbonyl amino methyl benzoic acid, benzoic acid, 4-1,1-dimethylethoxy carbonyl amino methyl | |
4-({[(tert-butoxy)carbonyl]amino}methyl)benzoate |
Specifications
168°C | |
White | |
MFCD00228182 | |
LNKHBRDWRIIROP-UHFFFAOYSA-M | |
4-({[(tert-butoxy)carbonyl]amino}methyl)benzoate | |
853605 | |
≥98.0% (HPLC,T) | |
Crystalline Powder |
33233-67-9 | |
C13H16NO4 | |
4-tert-butoxycarbonylamino methyl benzoic acid, 4-tert-butoxycarbonyl amino methyl benzoic acid, 4-tert-butoxy carbonyl amino methyl benzoic acid, boc-4-aminomethyl-benzoic acid, 4-boc-aminomethyl-benzoic acid, 4-tert-butoxycarbonylaminomethyl benzoic acid, 4-n-tert-butyloxycarbonyl amino methyl benzoic acid, benzoic acid, 4-1,1-dimethylethoxy carbonyl amino methyl | |
CC(C)(C)OC(=O)NCC1=CC=C(C=C1)C([O-])=O | |
250.28 | |
251.28 | |
1 g | |
4-[(tert-Butoxycarbonylamino)methyl]benzoic Acid |
Safety and Handling
TSCA : No