missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-tert-Butyl-3-methoxy-2,6-dinitrotoluene 99.0+%, TCI America™
Supplier: TCI America B1379500G
Specifications
4-tert-Butyl-3-methoxy-2,6-dinitrotoluene | |
86°C | |
185°C | |
MFCD00024268 | |
6-tert-Butyl-3-methyl-2,4-dinitroanisole | |
CC1=C(C=C(C(=C1[N+](=O)[O-])OC)C(C)(C)C)[N+](=O)[O-] | |
268.269 | |
CHEBI:82495 | |
≥99.0% (GC) |
83-66-9 | |
Yellow | |
C12H16N2O5 | |
500 g | |
SUAUILGSCPYJCS-UHFFFAOYSA-N | |
1-tert-butyl-2-methoxy-4-methyl-3,5-dinitrobenzene | |
6753 | |
268.27 | |
Crystalline Powder |
Chemical Identifiers
83-66-9 | |
268.269 | |
SUAUILGSCPYJCS-UHFFFAOYSA-N | |
6753 | |
1-tert-butyl-2-methoxy-4-methyl-3,5-dinitrobenzene |
C12H16N2O5 | |
MFCD00024268 | |
6-tert-Butyl-3-methyl-2,4-dinitroanisole | |
CHEBI:82495 | |
CC1=C(C=C(C(=C1[N+](=O)[O-])OC)C(C)(C)C)[N+](=O)[O-] |
Safety and Handling
RTECSNumber : BZ8575000
TSCA : Yes