Learn More
4-tert-Butylbenzenesulfonyl chloride, 98%, Thermo Scientific™
Chemical Identifiers
CAS | 15084-51-2 |
---|---|
Molecular Formula | C10H13ClO2S |
Molecular Weight (g/mol) | 232.73 |
MDL Number | MFCD00007449 |
InChI Key | YEZADZMMVHWFIY-UHFFFAOYSA-N |
Synonym | 4-t-butylbenzenesulfonyl chloride, 4-tert-butyl benzene-1-sulfonyl chloride, 4-tert-butylbenzene-1-sulfonyl chloride, 4-tert-butylbenzenesulfonylchloride, 4-tert-butyl benzenesulfonyl chloride, 4-tert-butyl-benzenesulfonyl chloride, 4-tert-butylphenylsulfonyl chloride, p-tert-butylbenzenesulfonyl chloride, 4-tert butylbenzenesulfonyl chloride, 4-tert-butyl phenylsulfonyl chloride |
PubChem CID | 139882 |
IUPAC Name | 4-tert-butylbenzenesulfonyl chloride |
SMILES | CC(C)(C)C1=CC=C(C=C1)S(=O)(=O)Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC303470100
|
Thermo Scientific Chemicals
303470100 |
10g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC303470500
|
Thermo Scientific Chemicals
303470500 |
50g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
15084-51-2 | |
232.73 | |
YEZADZMMVHWFIY-UHFFFAOYSA-N | |
139882 | |
CC(C)(C)C1=CC=C(C=C1)S(=O)(=O)Cl |
C10H13ClO2S | |
MFCD00007449 | |
4-t-butylbenzenesulfonyl chloride, 4-tert-butyl benzene-1-sulfonyl chloride, 4-tert-butylbenzene-1-sulfonyl chloride, 4-tert-butylbenzenesulfonylchloride, 4-tert-butyl benzenesulfonyl chloride, 4-tert-butyl-benzenesulfonyl chloride, 4-tert-butylphenylsulfonyl chloride, p-tert-butylbenzenesulfonyl chloride, 4-tert butylbenzenesulfonyl chloride, 4-tert-butyl phenylsulfonyl chloride | |
4-tert-butylbenzenesulfonyl chloride |
Specifications
15084-51-2 | |
100.0 | |
97.5% min. (GC) | |
C10H13ClO2S | |
MFCD00007449 | |
4-t-butylbenzenesulfonyl chloride, 4-tert-butyl benzene-1-sulfonyl chloride, 4-tert-butylbenzene-1-sulfonyl chloride, 4-tert-butylbenzenesulfonylchloride, 4-tert-butyl benzenesulfonyl chloride, 4-tert-butyl-benzenesulfonyl chloride, 4-tert-butylphenylsulfonyl chloride, p-tert-butylbenzenesulfonyl chloride, 4-tert butylbenzenesulfonyl chloride, 4-tert-butyl phenylsulfonyl chloride | |
CC(C)(C)C1=CC=C(C=C1)S(=O)(=O)Cl | |
232.73 | |
232.73 | |
4-tert-Butylbenzenesulfonyl chloride |
97.5 | |
Authentic | |
Glass bottle | |
(CH3)3CC6H4SO2Cl | |
10g | |
YEZADZMMVHWFIY-UHFFFAOYSA-N | |
4-tert-butylbenzenesulfonyl chloride | |
139882 | |
98% |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RUO â Research Use Only