Learn More
4-tert-Butylbenzoyl chloride, 98%, Thermo Scientific Chemicals
$112.60 - $331.97
Chemical Identifiers
CAS | 1710-98-1 |
---|---|
Molecular Formula | C11H13ClO |
Molecular Weight (g/mol) | 196.67 |
MDL Number | MFCD00000695 |
InChI Key | WNLMYNASWOULQY-UHFFFAOYSA-N |
PubChem CID | 74372 |
IUPAC Name | 4-tert-butylbenzoyl chloride |
SMILES | CC(C)(C)C1=CC=C(C=C1)C(Cl)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1437314
|
Thermo Scientific Chemicals
A1437314 |
25 g |
Each for $112.60
|
|
|||||
AAA1437322
|
Thermo Scientific Chemicals
A1437322 |
100 g |
Each for $331.97
|
|
|||||
Description
4-tert-Butylbenzoyl chloride is used in the synthesis of 9,9?-spirobifluorene-bridged bipolar systems containing 1,3,4-oxadiazole-conjugated oligoaryl and triaryllamine moieties. It is also used in the preparation of 2-(4-tert-butylphenyl)-5-(4-methoxyphenyl)-1,3,4-oxadiazole.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-tert-Butylbenzoyl chloride is used in the synthesis of 9,9′-spirobifluorene-bridged bipolar systems containing 1,3,4-oxadiazole-conjugated oligoaryl and triaryllamine moieties. It is also used in the preparation of 2-(4-tert-butylphenyl)-5-(4-methoxyphenyl)-1,3,4-oxadiazole.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from oxidizing agents, bases and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.
Chemical Identifiers
1710-98-1 | |
196.67 | |
WNLMYNASWOULQY-UHFFFAOYSA-N | |
4-tert-butylbenzoyl chloride |
C11H13ClO | |
MFCD00000695 | |
74372 | |
CC(C)(C)C1=CC=C(C=C1)C(Cl)=O |
Specifications
1710-98-1 | |
125°C to 126°C (10 mmHg) | |
C11H13ClO | |
MFCD00000695 | |
UN3265 | |
Moisture sensitive | |
Reacts with water. | |
CC(C)(C)C1=CC=C(C=C1)C(Cl)=O | |
196.67 | |
196.68 | |
4-tert-Butylbenzoyl chloride |
1.08 | |
87°C (188°F) | |
1.5351 | |
25 g | |
775793 | |
benzoyl chloride, 4-1,1-dimethylethyl, 4-tert-butylbenzoylchloride, p-t-butylbenzoyl chloride, p-tert-butylbenzoyl chloride, benzoyl chloride,4-1,1-dimethylethyl, 4-tert-butyl-benzoyl chloride, 4-tert-butyl benzoyl chloride, 4-t-butylbenzoyl chloride, acmc-1brh5, 4-tertbutylbenzoyl chloride | |
WNLMYNASWOULQY-UHFFFAOYSA-N | |
4-tert-butylbenzoyl chloride | |
74372 | |
98% |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302+H312+H332-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 216-973-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only